Merck
  • Differential activity of matrix metalloproteinases (MMPs) during photoperiod induced uterine regression and recrudescence in Siberian hamsters (Phodopus sungorus).
Differential activity of matrix metalloproteinases (MMPs) during photoperiod induced uterine regression and recrudescence in Siberian hamsters (Phodopus sungorus).
Molecular reproduction and development (2008-01-24)
Asha Shahed, Kelly A Young
ABSTRACT
Siberian hamsters adapt to seasonal changes by reducing their reproductive function during short days (SD). SD exposure reduces uterine mass and reproductive capacity, but underlying cellular mechanisms remain unknown. Because matrix metalloproteinases (MMPs) and their tissue inhibitors (TIMPs) are important in uterine development, parturition, and postpartum remodeling, their expression in uterine tissue from Siberian hamsters undergoing photoperiod-mediated reproductive regression and recrudescence was investigated. Female hamsters were exposed to long day (LD, 16L:8D, controls) or SD (8L:16D) for 3-12 weeks (regression); a second group was exposed to SD or LD for 14 weeks and then transferred to LD for 0-8 weeks (recrudescence). Hamsters were euthanized, uteri collected, and homogenates analyzed by gelatin zymography or Western blotting for MMP and TIMP protein levels. Uterine weight decreased (67-75%) at SD weeks 12-14 and increased post-LD transfer (PT) reaching LD values by PT week 2. MMP-2, but not MMP-9 activity was reduced by SD week 12 or 14 but increased to LD levels at PT week 2. MMP-3 expression increased at SD week 9 compared to other SD and LD groups. MMP-14 and -13 protein levels decreased at SD week 3 but returned to LD levels by SD week 6. During recrudescence, MMP-3 (PT weeks 0-2), MMP-13 (PT week 4), and MMP-14 (PT weeks 2, 4) protein levels were higher than LD. TIMP-1 and 2 were present at low levels. Significant and differential variations in uterine MMP activity/expression during photoperiod-induced regression and recrudescence were observed. These changes likely reflect increases in tissue remodeling during both the adaptation to SD and the restoration of reproductive function.
Sigma-Aldrich
Anti-MMP-14 Antibody, hemopexin domain, clone 113-5B7, clone 113-5B7, Chemicon®, from mouse
Sigma-Aldrich
Anti-TIMP-2 Antibody, clone 67-4H11, clone 67-4H11, Chemicon®, from mouse
Sigma-Aldrich
Anti-MMP-3 Antibody, clone SL-1 IID4, clone SL-1 IID4, Chemicon®, from mouse
Sigma-Aldrich
Anti-MMP-13 Antibody, clone 181-15A12, clone 181-15A12, Chemicon®, from mouse