Merck
  • Astragaloside IV attenuates high glucose-induced NF-κB-mediated inflammation through activation of PI3K/AKT-ERK-dependent Nrf2/ARE signaling pathway in glomerular mesangial cells.
Astragaloside IV attenuates high glucose-induced NF-κB-mediated inflammation through activation of PI3K/AKT-ERK-dependent Nrf2/ARE signaling pathway in glomerular mesangial cells.
Phytotherapy research : PTR (2023-05-16)
Xue Su, Hengjiang Guo, Yuying Zhou, Aili Cao, Qian Shen, Bingbing Zhu, Xingmei Yao, Yunman Wang, Hao Wang, Li Wang
ABSTRACT
Inflammation is a key contributor to diabetic kidney disease pathogenesis, including reactive oxidation stress (ROS)-mediated nuclear factor-κB (NF-κB) signaling pathway. In this study, we examined the effect of Astragaloside IV (AS-IV) on anti-inflammatory and anti-oxidative properties under high glucose (HG) condition and the potential mechanism in glomerular mesangial cells (GMCs). We showed that AS-IV concentration-dependently reduced GMCs proliferation, restrained ROS release and hydrogen peroxide content, and suppressed pro-inflammatory cytokines as well as pro-fibrotic factors expression, which were associated with the inhibition of NF-κB and nuclear factor-erythroid 2-related factor 2 (Nrf2) signaling activation. Accordingly, both NF-κB overexpression by using RNA plasmid and Nrf2 gene silencing by using RNA interference weakened the ability of AS-IV to ameliorate HG-induced oxidative stress, inflammation, and cell proliferation. Furthermore, phosphatidylinositide 3-kinases (PI3K)/serine/threonine protein kinase (Akt) and extracellular regulated protein kinases (ERK) signaling pathway regulated the process of AS-IV-induced Nrf2 activation and antioxidant capacity, which evidenced by using PI3K inhibitor LY294002 or ERK inhibitor PD98059 that largely abolished the AS-IV efficacy. Taken together, these results indicated that AS-IV protected against HG-induced GMCs damage by inhibiting ROS/NF-kB-induced increases of inflammatory cytokines, fibrosis biomarkers, and cell proliferation via up-regulation of Nrf2-dependent antioxidant enzyme expression, which were mediated by PI3K/Akt and ERK signaling pathway activation.
Sigma-Aldrich
Monoclonal Anti-MAP Kinase, Activated (Diphosphorylated ERK-1&2) antibody produced in mouse, clone MAPK-YT, ascites fluid
Sigma-Aldrich
Anti-p38 MAP Kinase, Activated (Diphosphorylated p38) antibody, Mouse monoclonal, clone P38-TY, purified from hybridoma cell culture, buffered aqueous solution
Sigma-Aldrich
Anti-phospho-p38 MAPK (pTyr182) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-phospho-Erk1/2 (Thr202/Tyr204, Thr185/Tyr187)Antibody, recombinant clone AW39R, rabbit monoclonal, clone AW39R, Upstate®, from rabbit