Merck
Cellular Senescence Is Immunogenic and Promotes Antitumor Immunity.
Cancer discovery (2022-10-28)
Ines Marin, Olga Boix, Andrea Garcia-Garijo, Isabelle Sirois, Adrià Caballe, Eduardo Zarzuela, Irene Ruano, Camille Stephan-Otto Attolini, Neus Prats, José A López-Domínguez, Marta Kovatcheva, Elena Garralda, Javier Muñoz, Etienne Caron, María Abad, Alena Gros, Federico Pietrocola, Manuel Serrano
ABSTRACT
Cellular senescence is a stress response that activates innate immune cells, but little is known about its interplay with the adaptive immune system. Here, we show that senescent cells combine several features that render them highly efficient in activating dendritic cells (DC) and antigen-specific CD8 T cells. This includes the release of alarmins, activation of IFN signaling, enhanced MHC class I machinery, and presentation of senescence-associated self-peptides that can activate CD8 T cells. In the context of cancer, immunization with senescent cancer cells elicits strong antitumor protection mediated by DCs and CD8 T cells. Interestingly, this protection is superior to immunization with cancer cells undergoing immunogenic cell death. Finally, the induction of senescence in human primary cancer cells also augments their ability to activate autologous antigen-specific tumor-infiltrating CD8 lymphocytes. Our study indicates that senescent cancer cells can be exploited to develop efficient and protective CD8-dependent antitumor immune responses. Our study shows that senescent cells are endowed with a high immunogenic potential-superior to the gold standard of immunogenic cell death. We harness these properties of senescent cells to trigger efficient and protective CD8-dependent antitumor immune responses. See related article by Chen et al., p. 432. This article is highlighted in the In This Issue feature, p. 247.
Sigma-Aldrich
Bleomycin sulfate from Streptomyces verticillus, 1.5-2.0 units/mg solid, BioReagent, suitable for cell culture
Sigma-Aldrich
Nutlin-3a, ≥98% (HPLC)