Merck
  • Astrocyte-oligodendrocyte interaction regulates central nervous system regeneration.
Astrocyte-oligodendrocyte interaction regulates central nervous system regeneration.
Nature communications (2023-06-09)
Irene Molina-Gonzalez, Rebecca K Holloway, Zoeb Jiwaji, Owen Dando, Sarah A Kent, Katie Emelianova, Amy F Lloyd, Lindsey H Forbes, Ayisha Mahmood, Thomas Skripuletz, Viktoria Gudi, James A Febery, Jeffrey A Johnson, Jill H Fowler, Tanja Kuhlmann, Anna Williams, Siddharthan Chandran, Martin Stangel, Andrew J M Howden, Giles E Hardingham, Veronique E Miron
ABSTRACT
Failed regeneration of myelin around neuronal axons following central nervous system damage contributes to nerve dysfunction and clinical decline in various neurological conditions, for which there is an unmet therapeutic demand. Here, we show that interaction between glial cells - astrocytes and mature myelin-forming oligodendrocytes - is a determinant of remyelination. Using in vivo/ ex vivo/ in vitro rodent models, unbiased RNA sequencing, functional manipulation, and human brain lesion analyses, we discover that astrocytes support the survival of regenerating oligodendrocytes, via downregulation of the Nrf2 pathway associated with increased astrocytic cholesterol biosynthesis pathway activation. Remyelination fails following sustained astrocytic Nrf2 activation in focally-lesioned male mice yet is restored by either cholesterol biosynthesis/efflux stimulation, or Nrf2 inhibition using the existing therapeutic Luteolin. We identify that astrocyte-oligodendrocyte interaction regulates remyelination, and reveal a drug strategy for central nervous system regeneration centred on targeting this interaction.
Sigma-Aldrich
Anti-Olig2 Antibody, clone 211F1.1, clone 211F1.1, from mouse
Sigma-Aldrich
Anti-Vimentin Antibody, clone LN-6, culture supernatant, clone LN-6, Chemicon®
Sigma-Aldrich
Anti-Sox9 Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Anti-Ki-67 Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Anti-Myelin Associated Glycoprotein Antibody, clone 513, clone 513, Chemicon®, from mouse
Sigma-Aldrich
Anti-Olig1 Antibody, ascites fluid, Chemicon®
Sigma-Aldrich
Anti-Olig-2 Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Monoclonal Anti-SOX10 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL4455, purified immunoglobulin
Sigma-Aldrich
Monoclonal Anti-CNP antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL2887, purified immunoglobulin, buffered aqueous glycerol solution