Merck
  • Airway surface hyperviscosity and defective mucociliary transport by IL-17/TNF-α are corrected by beta-adrenergic stimulus.
Airway surface hyperviscosity and defective mucociliary transport by IL-17/TNF-α are corrected by beta-adrenergic stimulus.
JCI insight (2022-10-12)
Daniela Guidone, Martina Buccirossi, Paolo Scudieri, Michele Genovese, Sergio Sarnataro, Rossella De Cegli, Federico Cresta, Vito Terlizzi, Gabrielle Planelles, Gilles Crambert, Isabelle Sermet-Gaudelus, Luis Jv Galietta
ABSTRACT
The fluid covering the surface of airway epithelia represents a first barrier against pathogens. The chemical and physical properties of the airway surface fluid are controlled by the activity of ion channels and transporters. In cystic fibrosis (CF), loss of CFTR chloride channel function causes airway surface dehydration, bacterial infection, and inflammation. We investigated the effects of IL-17A plus TNF-α, two cytokines with a relevant role in CF and other chronic lung diseases. Transcriptome analysis revealed a profound change with upregulation of several genes involved in ion transport, anti-bacterial defense, and neutrophil recruitment. At the functional level, bronchial epithelia treated in vitro with the cytokine combination showed upregulation of ENaC sodium channel, ATP12A proton pump, ADRB2 beta-adrenergic receptor, and SLC26A4 anion exchanger. The overall result of IL-17A/TNF-α treatment was hyperviscosity of the airway surface as demonstrated by fluorescence recovery after photobleaching (FRAP) experiments. Importantly, stimulation with a beta-adrenergic agonist switched airway surface to a low viscosity state in non-CF but not in CF epithelia. Our study suggests that CF lung disease is sustained by a vicious cycle in which epithelia cannot exit from the hyperviscous state thus perpetuating the proinflammatory airway surface condition.
Sigma-Aldrich
Interleukin-1β human, IL-1β, recombinant, expressed in HEK 293 cells, HumanKine®, suitable for cell culture, endotoxin tested
Sigma-Aldrich
Anti-Acetylated Tubulin antibody, Mouse monoclonal, clone 6-11B-1, purified from hybridoma cell culture
Sigma-Aldrich
Interleukin-6 human, IL-6, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, protease free, fatty acid free, essentially globulin free, pH 7, ≥98%
Sigma-Aldrich
Interferon-γ human, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), lyophilized powder, suitable for cell culture
Sigma-Aldrich
Anti-SCNN1B antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Interferon α 2A human, IFN-α 2A, recombinant, expressed in HEK 293 cells, HumanKine®, suitable for cell culture
Sigma-Aldrich
IL-17 human, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
Anti-SCNN1A antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Tumor Necrosis Factor-α human, TNF-α, recombinant, expressed in E. coli, powder, suitable for cell culture
Sigma-Aldrich
Anti-Glyceraldehyde-3-Phosphate Dehydrogenase Antibody, clone 6C5, clone 6C5, Chemicon®, from mouse