Merck
  • A tautomeric ligand enables directed C‒H hydroxylation with molecular oxygen.
A tautomeric ligand enables directed C‒H hydroxylation with molecular oxygen.
Science (New York, N.Y.) (2021-11-30)
Zhen Li, Zhen Wang, Nikita Chekshin, Shaoqun Qian, Jennifer X Qiao, Peter T Cheng, Kap-Sun Yeung, William R Ewing, Jin-Quan Yu
ABSTRACT
Hydroxylation of aryl carbon-hydrogen bonds with transition metal catalysts has proven challenging when oxygen is used as the oxidant. Here, we report a palladium complex bearing a bidentate pyridine/pyridone ligand that efficiently catalyzes this reaction at ring positions adjacent to carboxylic acids. Infrared, x-ray, and computational analysis support a possible role of ligand tautomerization from mono-anionic (L,X) to neutral (L,L) coordination in the catalytic cycle of aerobic carbon-hydrogen hydroxylation reaction. The conventional site selectivity dictated by heterocycles is overturned by this catalyst, thus allowing late-stage modification of compounds of pharmaceutical interest at previously inaccessible sites.
Sigma-Aldrich
6-(2-(5-chloropyridin-2-yl)-1-phenylpropan-2-yl)pyridin-2-ol
Sigma-Aldrich
2-fluoro-6-(pyrimidin-5-yl)aniline, ≥95%
Sigma-Aldrich
6-(1-Cyclohexyl-1-(5-methylpyridin-2-yl)ethyl)pyridin-2(1H)-one, ≥95%
Sigma-Aldrich
6-(2-(Quinolin-2-yl)propan-2-yl)pyridin-2-ol, ≥95%
Sigma-Aldrich
6-(Quinolin-2-yl)pyridin-2(1H)-one, ≥95%