Merck
  • Leptin Induces Apoptotic and Pyroptotic Cell Death via NLRP3 Inflammasome Activation in Rat Hepatocytes.
Leptin Induces Apoptotic and Pyroptotic Cell Death via NLRP3 Inflammasome Activation in Rat Hepatocytes.
International journal of molecular sciences (2021-11-28)
Ananda Baral, Pil-Hoon Park
ABSTRACT
Leptin, a hormone that is predominantly produced by adipose tissue, is closely associated with various liver diseases. However, there is a lack of understanding as to whether leptin directly induces cytotoxic effects in hepatocytes as well as the mechanisms that are involved. Inflammasomes, which are critical components in the innate immune system, have been recently shown to modulate cell death. In this study, we examined the effect of leptin on the viability of rat hepatocytes and the underlying mechanisms, with a particular focus on the role of inflammasomes activation. Leptin treatment induced cytotoxicity in rat hepatocytes, as determined by decreased cell viability, increased caspase-3 activity, and the enhanced release of lactate dehydrogenase. NLRP3 inflammasomes were activated by leptin both in vitro and in vivo, as determined by the maturation of interleukin-1β and caspase-1, and the increased expression of inflammasome components, including NLRP3 and ASC. Mechanistically, leptin-induced inflammasome activation is mediated via the axis of ROS production, ER stress, and autophagy. Notably, the inhibition of inflammasomes by treatment with the NLRP3 inhibitor or the IL-1 receptor antagonist protected the hepatocytes from leptin-induced cell death. Together, these results indicate that leptin exerts cytotoxic effects in hepatocytes, at least in part, via the activation of NLRP3 inflammasomes.
Sigma-Aldrich
Collagen, Type I solution from rat tail, BioReagent, suitable for cell culture, sterile-filtered
Sigma-Aldrich
3-Methyladenine, autophagy inhibitor
Sigma-Aldrich
Collagenase from Clostridium histolyticum, suitable for release of physiologically active rat hepatocytes, Type IV, 0.5-5.0 FALGPA units/mg solid, ≥125 CDU/mg solid
Sigma-Aldrich
Ac-YVAD-cmk, ≥95% (HPLC)
Sigma-Aldrich
N-Acetyl-L-cysteine, BioReagent, suitable for cell culture
Sigma-Aldrich
Leptin from mouse, ≥98% (SDS-PAGE), recombinant, expressed in E. coli, lyophilized powder
Sigma-Aldrich
CA-074, CA-074, also referenced under CAS 134448-10-5, modulates the biological activity of cathepsin B. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.