Merck
  • Generation of mitochondrial reactive oxygen species is controlled by ATPase inhibitory factor 1 and regulates cognition.
Generation of mitochondrial reactive oxygen species is controlled by ATPase inhibitory factor 1 and regulates cognition.
PLoS biology (2021-05-14)
Pau B Esparza-Moltó, Inés Romero-Carramiñana, Cristina Núñez de Arenas, Marta P Pereira, Noelia Blanco, Beatriz Pardo, Georgina R Bates, Carla Sánchez-Castillo, Rafael Artuch, Michael P Murphy, José A Esteban, José M Cuezva
ABSTRACT
The mitochondrial ATP synthase emerges as key hub of cellular functions controlling the production of ATP, cellular signaling, and fate. It is regulated by the ATPase inhibitory factor 1 (IF1), which is highly abundant in neurons. Herein, we ablated or overexpressed IF1 in mouse neurons to show that IF1 dose defines the fraction of active/inactive enzyme in vivo, thereby controlling mitochondrial function and the production of mitochondrial reactive oxygen species (mtROS). Transcriptomic, proteomic, and metabolomic analyses indicate that IF1 dose regulates mitochondrial metabolism, synaptic function, and cognition. Ablation of IF1 impairs memory, whereas synaptic transmission and learning are enhanced by IF1 overexpression. Mechanistically, quenching the IF1-mediated increase in mtROS production in mice overexpressing IF1 reduces the increased synaptic transmission and obliterates the learning advantage afforded by the higher IF1 content. Overall, IF1 plays a key role in neuronal function by regulating the fraction of ATP synthase responsible for mitohormetic mtROS signaling.
Sigma-Aldrich
Anti-KEAP1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-NR2A Antibody, Upstate®, from rabbit
Sigma-Aldrich
Anti-NMDAR1 Antibody, rabbit monoclonal, culture supernatant, clone 1.17.2.6, Chemicon®
Sigma-Aldrich
Anti-CaM Kinase IIα (CaMKIIα) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone DM1A, ascites fluid
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, ascites fluid
Sigma-Aldrich
Anti-phospho-CaM Kinase II Antibody, α subunit, (Thr286), clone 22B1, clone 22B1, Upstate®, from mouse
Sigma-Aldrich
Anti-Glutamate Receptor 1 Antibody, phosphoSer 831, Chemicon®, from rabbit