Merck
  • Targeting KRAS4A splicing through the RBM39/DCAF15 pathway inhibits cancer stem cells.
Targeting KRAS4A splicing through the RBM39/DCAF15 pathway inhibits cancer stem cells.
Nature communications (2021-07-15)
Wei-Ching Chen, Minh D To, Peter M K Westcott, Reyno Delrosario, Il-Jin Kim, Mark Philips, Quan Tran, Saumya R Bollam, Hani Goodarzi, Nora Bayani, Olga Mirzoeva, Allan Balmain
ABSTRACT
The commonly mutated human KRAS oncogene encodes two distinct KRAS4A and KRAS4B proteins generated by differential splicing. We demonstrate here that coordinated regulation of both isoforms through control of splicing is essential for development of Kras mutant tumors. The minor KRAS4A isoform is enriched in cancer stem-like cells, where it responds to hypoxia, while the major KRAS4B is induced by ER stress. KRAS4A splicing is controlled by the DCAF15/RBM39 pathway, and deletion of KRAS4A or pharmacological inhibition of RBM39 using Indisulam leads to inhibition of cancer stem cells. Our data identify existing clinical drugs that target KRAS4A splicing, and suggest that levels of the minor KRAS4A isoform in human tumors can be a biomarker of sensitivity to some existing cancer therapeutics.
Sigma-Aldrich
Monoclonal Anti-RNPC2 antibody produced in mouse, clone 4G8, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
bisBenzimide H 33342 trihydrochloride, ≥98% (HPLC and TLC)
Sigma-Aldrich
Monoclonal Anti-KRAS antibody produced in mouse, clone 3B10-2F2, purified immunoglobulin, buffered aqueous solution