Merck
  • Siglec-1 Macrophages and the Contribution of IFN to the Development of Autoimmune Congenital Heart Block.
Siglec-1 Macrophages and the Contribution of IFN to the Development of Autoimmune Congenital Heart Block.
Journal of immunology (Baltimore, Md. : 1950) (2018-12-07)
Robert M Clancy, Marc Halushka, Sara E Rasmussen, Tenzin Lhakhang, Miao Chang, Jill P Buyon
ABSTRACT
Given that diseases associated with anti-SSA/Ro autoantibodies, such as systemic lupus erythematosus and Sjögren syndrome, are linked with an upregulation of IFN and type I IFN-stimulated genes, including sialic acid-binding Ig-like lectin 1 (Siglec-1), a receptor on monocytes/macrophages, recent attention has focused on a potential role for IFN and IFN-stimulated genes in the pathogenesis of congenital heart block (CHB). Accordingly, three approaches were leveraged to address the association of IFN, IFN-stimulated genes, and the phenotype of macrophages in affected fetal cardiac tissue: 1) cultured healthy human macrophages transfected with hY3, an anti-SSA/Ro-associated ssRNA, 2) RNA isolated from freshly sorted human leukocytes/macrophages after Langendorff perfusion of three fetal hearts dying with CHB and three healthy gestational age-matched hearts, and 3) autopsy tissue from three additional human CHB hearts and one healthy heart. TLR ligation of macrophages with hY3 led to the upregulation of a panel of IFN transcripts, including SIGLEC1, a result corroborated using quantitative PCR. Using independent and agnostic bioinformatics approaches, CD45+CD11c+ and CD45+CD11c- human leukocytes flow sorted from the CHB hearts highly expressed type I IFN response genes inclusive of SIGLEC1. Furthermore, Siglec-1 expression was identified in the septal region of several affected fetal hearts. These data now provide a link between IFN, IFN-stimulated genes, and the inflammatory and possibly fibrosing components of CHB, positioning Siglec-1-positive macrophages as integral to the process.
Sigma-Aldrich
Anti-SIGLEC1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous glycerol solution