Merck
Reduced RNA turnover as a driver of cellular senescence.
Life science alliance (2021-01-16)
Nowsheen Mullani, Yevheniia Porozhan, Adèle Mangelinck, Christophe Rachez, Mickael Costallat, Eric Batsché, Michele Goodhardt, Giovanni Cenci, Carl Mann, Christian Muchardt
ABSTRACT
Accumulation of senescent cells is an important contributor to chronic inflammation upon aging. The inflammatory phenotype of senescent cells was previously shown to be driven by cytoplasmic DNA. Here, we propose that cytoplasmic double-stranded RNA has a similar effect. We find that several cell types driven into senescence by different routes share an accumulation of long promoter RNAs and 3' gene extensions rich in retrotransposon sequences. Accordingly, these cells display increased expression of genes involved in response to double stranded RNA of viral origin downstream of the interferon pathway. The RNA accumulation is associated with evidence of reduced RNA turnover, including in some cases, reduced expression of RNA exosome subunits. Reciprocally, depletion of RNA exosome subunit EXOSC3 accelerated expression of multiple senescence markers. A senescence-like RNA accumulation was also observed in cells exposed to oxidative stress, an important trigger of cellular senescence. Altogether, we propose that in a subset of senescent cells, repeat-containing transcripts stabilized by oxidative stress or reduced RNA exosome activity participate in driving and maintaining the permanent inflammatory state characterizing cellular senescence.
Sigma-Aldrich
Doxycycline hydrochloride
Sigma-Aldrich
Anti-Actin, N-terminal antibody produced in rabbit, ~0.5 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
(Z)-4-Hydroxytamoxifen, ≥98% Z isomer
Sigma-Aldrich
ATX Ponceau S red staining solution, BioReagent, ready-to-use solution
Sigma-Aldrich
TWEEN® 20, viscous liquid
Sigma-Aldrich
MISSION® esiRNA, targeting human THPO