Merck
  • Determination of plasma levels of the active thiol form of the direct-acting PrC-210 ROS-scavenger using a fluorescence-based assay.
Determination of plasma levels of the active thiol form of the direct-acting PrC-210 ROS-scavenger using a fluorescence-based assay.
Analytical biochemistry (2021-01-09)
Emma Dreischmeier, William E Fahl
ABSTRACT
PrC-210 is a direct-acting ROS-scavenger. It's active when administered orally, IV, or topically; it has none of the nausea/emesis nor hypotension side effects that have precluded human amifostine use. PrC-210 confers 100% survival to mice and rats that received an otherwise 100% lethal radiation dose and 36% reduction of ischemia-reperfusion-induced mouse myocardial infarct damage, and thus is a viable candidate to prevent human ROS-induced ischemia-reperfusion and ionizing radiation toxicities. We report the first assay for the pharmacologically active PrC-210 thiol in blood. PrC-210 has no double-bonds nor light absorption, so derivatizing the thiol with a UV-absorbing fluorochrome enables quantification. This assay: i) is done on the benchtop; it's read with a fluorescence plate reader, ii) provides linear product formation through 60 min, iii) quantifies μM to low mM rodent blood levels of PrC-210 that confer complete radioprotection, iv) accurately reflects PrC-210 thiol formation of mixed disulfides with other thiols in blood, and v) shows excellent between-day assay outcome with very low standard deviation and coefficient of variation. A fluorescence assay quantifying formation of a PrC-210 thiol-bimane adduct enables measurement of blood PrC-210 thiol. A blood assay will help in the development of PrC-210 for use in the human clinical setting.
Sigma-Aldrich
Monochlorobimane, suitable for fluorescence, ≥70.0% (HPCE)
Sigma-Aldrich
DL-Cysteine, technical grade
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
Chlorobimane
SAFC
L-Cysteine