Merck
  • Gut bacteria-derived 5-hydroxyindole is a potent stimulant of intestinal motility via its action on L-type calcium channels.
Gut bacteria-derived 5-hydroxyindole is a potent stimulant of intestinal motility via its action on L-type calcium channels.
PLoS biology (2021-01-23)
Barbora Waclawiková, Amber Bullock, Markus Schwalbe, Carmen Aranzamendi, Sieger A Nelemans, Gertjan van Dijk, Sahar El Aidy
ABSTRACT
Microbial conversion of dietary or drug substrates into small bioactive molecules represents a regulatory mechanism by which the gut microbiota alters intestinal physiology. Here, we show that a wide variety of gut bacteria can metabolize the dietary supplement and antidepressant 5-hydroxytryptophan (5-HTP) to 5-hydroxyindole (5-HI) via the tryptophanase (TnaA) enzyme. Oral administration of 5-HTP results in detection of 5-HI in fecal samples of healthy volunteers with interindividual variation. The production of 5-HI is inhibited upon pH reduction in in vitro studies. When administered orally in rats, 5-HI significantly accelerates the total gut transit time (TGTT). Deciphering the underlying mechanisms of action reveals that 5-HI accelerates gut contractility via activation of L-type calcium channels located on the colonic smooth muscle cells. Moreover, 5-HI stimulation of a cell line model of intestinal enterochromaffin cells results in significant increase in serotonin production. Together, our findings support a role for bacterial metabolism in altering gut motility and lay the foundation for microbiota-targeted interventions.
Product Description
Millipore
Kovac′s reagent for indoles, suitable for microbiology
Sigma-Aldrich
Ondansetron hydrochloride dihydrate, ≥98% (HPLC), powder
Sigma-Aldrich
Fluoxetine hydrochloride, solid
Sigma-Aldrich
Acetylcholine chloride, suitable for cell culture
Sigma-Aldrich
5-Hydroxyindole, 97%
Sigma-Aldrich
Serotonin hydrochloride, powder
Sigma-Aldrich
SB-207266, ≥98% (HPLC)
Roche
DNase I, from bovine pancreas
Sigma-Aldrich
Pyridoxal 5′-phosphate hydrate, ≥98%
Sigma-Aldrich
5-Hydroxy-L-tryptophan, powder
Sigma-Aldrich
Carmine, powder, ≥42.0% (UV-vis)
Sigma-Aldrich
Methyl cellulose, viscosity: 4,000 cP