Merck
  • Comparison of Biochemical Constituents and Contents in Floral Nectar of Castanea spp.
Comparison of Biochemical Constituents and Contents in Floral Nectar of Castanea spp.
Molecules (Basel, Switzerland) (2020-09-19)
Young Ki Kim, Sujin Lee, Jeong Ho Song, Mahn Jo Kim, Ural Yunusbaev, Myeong-Lyeol Lee, Mun Seop Kim, Hyung Wook Kwon
ABSTRACT
Pollination is essential for efficient reproduction in pollinator-dependent crops that rely on the attraction of pollinators to flowers. Especially, floral nectar is considered to be an important factor attracting pollinator like honey bees, but differences among major chestnut species (Castanea crenata, C. mollissima, C. dentata, and C. sativa) are still little explored. This study aims to evaluate the value of honey source by analyzing floral nectar characteristics and comparing the composition of volatile organic compounds (VOCs) that mediate plant-pollinator interaction. In this study, we analyzed nectar samples obtained from male flowers using HPLC and HS-SPME/GC-MS. The five chestnuts showed significant differences between the volume of secreted nectar, free sugar composition, amino acid content and VOCs composition. Furthermore, C. crenata (Japanese cultivar 'Ungi') was revealed to emit the highest total amounts of VOCs and high levels of benzenoid compounds that are generally associated with flower-visiting insects. The sugar content per catkin, which is used to determine the honey yield, was the highest in C. crenata, suggesting that C. crenata 'Ungi' can be highly valued as a honey tree. Therefore, a better understanding of the relationship between pollinator and nectar characteristics of C. crenara could contribute to a prospective honey plant.
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-(−)-Fructose, BioUltra, ≥99.0% (HPLC)
Supelco
Alkane standard solution C8-C20, analytical standard, contains C8-C20, ~40 mg/L each, in hexane
Supelco
Acetophenone, analytical standard
Sigma-Aldrich
Sucrose, BioUltra, for molecular biology, ≥99.5% (HPLC)