Merck
  • Ultrasound Microbubble-Mediated VHL Regulates the Biological Behavior of Ovarian Cancer Cells.
Ultrasound Microbubble-Mediated VHL Regulates the Biological Behavior of Ovarian Cancer Cells.
Ultrasound in medicine & biology (2020-12-03)
Cong Li, Suling Hu, Yan Yue
ABSTRACT
According to the literature, the von Hippel-Lindau (VHL) gene has a certain correlation with ovarian cancer. In this study, we investigated the effect and mechanism of ultrasound microbubble-mediated VHL on the biological function of ovarian cancer cells. Non-targeting lipid microbubbles and targeted lipid microbubbles were prepared. OVCAR-3 cells were treated with VHL mediated by ultrasound and microbubbles alone or together. Expressions of VHL, Akt, epithelial-mesenchymal-transition-related proteins and apoptosis-related proteins were detected by Western blot and quantitative real-time polymerase chain reaction as needed. The effect of ultrasound microbubble-mediated VHL on the proliferation, apoptosis, cell cycle, migration and invasion of OVCAR-3 cells was examined by Cell Counting Kit-8, flow cytometry, wound-healing assay and Transwell. Compared with other treatment methods, ultrasound microbubble mediation enhanced VHL expression in OVCAR-3 cells. Overexpression of liposome-mediated VHL inhibited the proliferation and migration; caused cell-cycle arrest; promoted apoptosis: downregulated the expressions of MMP2, MMP9, E-cadherin, Akt and Bcl-2; and upregulated the expressions of VHL and BCL2-associated X protein (BAX) in OVCAR-3 cells. The effect of microbubble-treated VHL was similar to liposome-mediated regulation, while ultrasound treatment enhanced the effect of VHL on OVCAR-3 cells. More interestingly, ultrasound microbubble-mediated VHL had the most obvious regulatory effect on OVCAR-3 cells. Ultrasound microbubble technology increases the transfection efficiency of VHL into OVCAR-3 cells and enhances the effect of VHL gene on the biological function of OVCAR-3 cells.
Sigma-Aldrich
1,2-Distearoyl-sn-glycero-3-phosphoethanolamine, ≥99%
SKU
Pack Size
Availability
Price
Quantity
Corning® Transwell® polyester membrane cell culture inserts, 24 mm Transwell with 0.4 μm pore polyester membrane insert, TC-treated, sterile, 24/cs
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
1,2-Dipalmitoyl-sn-glycero-3-phosphate sodium salt, ≥99%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Glycerol, for molecular biology, ≥99.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Phosphate buffered saline, tablet