Merck
  • Sustained hedgehog signaling in medulloblastoma tumoroids is attributed to stromal astrocytes and astrocyte-derived extracellular matrix.
Sustained hedgehog signaling in medulloblastoma tumoroids is attributed to stromal astrocytes and astrocyte-derived extracellular matrix.
Laboratory investigation; a journal of technical methods and pathology (2020-05-28)
Yan Cheng, Janusz Franco-Barraza, Yuan Wang, Chaonan Zheng, Li Zhang, Yanghui Qu, Yaqiu Long, Edna Cukierman, Zeng-Jie Yang
ABSTRACT
Aberrant activation of the hedgehog (Hh) signaling pathway is associated with the formation of medulloblastoma (MB), the most common malignant pediatric brain tumor. However, tumor cells from human and mouse MB can not be passaged or preserved after being adherently cultured. Moreover, Hh signaling in MB cells is inactivated in such culture. Here we demonstrate that MB cells are capable of forming tumoroids (tumor spheroids) in vitro under optimized conditions, which can be further passaged and cryopreserved. More importantly, MB cells maintain Hh pathway activation and cell proliferation in tumoroids. Our studies further reveal that tumoroids-forming capacity of MB cells relies on astrocytes, a major component of the MB microenvironment. Astrocytes facilitate the formation of MB tumoroids by secreting sonic hedgehog (Shh) and generating astrocyte-derived extracellular matrix. These findings demonstrate the critical role of stromal astrocytes in supporting the survival and proliferation of MB cells in vitro. This study establishes a valid model for long-term culture of primary MB cells, which could be greatly beneficial for future investigation of MB tumorigenicity and the development of improved approaches to treat MB.
Sigma-Aldrich
Anti-Integrin α5β1 Antibody, clone BMA5, culture supernatant, clone BMA5, from rat
Sigma-Aldrich
Rat IgG1 Negative Control, clone HRPN, Azide Free Antibody, clone HRPN, from rat, purified by affinity chromatography
Sigma-Aldrich
Anti-Integrin Beta1, clone AIIB2 (Azide Free) Antibody, clone AIIB2, 1 mg/mL, from rat
Sigma-Aldrich
Rat IgG2b Negative Control, clone LTF-2, Azide Free Antibody, clone LTF-2, from rat, purified by affinity chromatography