Merck
Confocal Fluorescence-Lifetime Single-Molecule Localization Microscopy.
ACS nano (2020-10-10)
Jan Christoph Thiele, Dominic A Helmerich, Nazar Oleksiievets, Roman Tsukanov, Eugenia Butkevich, Markus Sauer, Oleksii Nevskyi, Jörg Enderlein
ABSTRACT
Fluorescence lifetime imaging microscopy is an important technique that adds another dimension to intensity and color acquired by conventional microscopy. In particular, it allows for multiplexing fluorescent labels that have otherwise similar spectral properties. Currently, the only super-resolution technique that is capable of recording super-resolved images with lifetime information is stimulated emission depletion microscopy. In contrast, all single-molecule localization microscopy (SMLM) techniques that employ wide-field cameras completely lack the lifetime dimension. Here, we combine fluorescence-lifetime confocal laser-scanning microscopy with SMLM for realizing single-molecule localization-based fluorescence-lifetime super-resolution imaging. Besides yielding images with a spatial resolution much beyond the diffraction limit, it determines the fluorescence lifetime of all localized molecules. We validate our technique by applying it to direct stochastic optical reconstruction microscopy and points accumulation for imaging in nanoscale topography imaging of fixed cells, and we demonstrate its multiplexing capability on samples with two different labels that differ only by fluorescence lifetime but not by their spectral properties.
Sigma-Aldrich
Anti-β-Tubulin antibody, Mouse monoclonal, ~2.0 mg/mL, clone AA2, purified from hybridoma cell culture
Sigma-Aldrich
Anti-α-Tubulin antibody, Mouse monoclonal, clone DM1A, purified from hybridoma cell culture
Sigma-Aldrich
Sodium borohydride, purum p.a., ≥96% (gas-volumetric)
Sigma-Aldrich
Magnesium chloride hexahydrate, ACS reagent, 99.0-102.0%
Sigma-Aldrich
Anti-Mouse IgG (H+L), highly cross adsorbed antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
Sigma-Aldrich
MES hydrate, ≥99.5% (titration)
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
Dulbecco′s Phosphate Buffered Saline, Modified, without calcium chloride and magnesium chloride, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, BioUltra, for molecular biology, ≥99.0% (T)