Merck
  • Structure-Based Scaffold Repurposing toward the Discovery of Novel Cholinesterase Inhibitors.
Structure-Based Scaffold Repurposing toward the Discovery of Novel Cholinesterase Inhibitors.
ACS omega (2020-12-17)
Satish N Dighe, Mangapathiraju Tippana, Suzannah van Akker, Trudi A Collet
ABSTRACT
Cholinesterases (ChE) are well-known drug targets for the treatment of Alzheimer's disease (AD). In continuation of work to develop novel cholinesterase inhibitors, we utilized a structure-based scaffold repurposing approach and discovered six novel ChE inhibitors from our recently developed DNA gyrase inhibitor library. Among the identified hits, two compounds (denoted 3 and 18) were found to be the most potent inhibitor of acetylcholinesterase (AChE, IC50 = 6.10 ± 1.01 μM) and butyrylcholinesterase (BuChE, IC50 = 5.50 ± 0.007 μM), respectively. Compound 3 was responsible for the formation of H-bond and π-π stacking interactions within the active site of AChE. In contrast, compound 18 was well fitted in the choline-binding pocket and catalytic site of BuChE. Results obtained from in vitro cytotoxicity assays and in silico derived physicochemical and absorption, distribution, metabolism, and excretion (ADME) properties indicate that repurposed scaffold 3 and 18 could be potential drug candidates for further development as novel ChE inhibitors.
Sigma-Aldrich
Acetylthiocholine iodide, ≥99.0% (AT)
Supelco
Trizma® hydrochloride buffer solution, BioUltra, pH 7.4
Sigma-Aldrich
Acetylcholinesterase from Electrophorus electricus (electric eel), Type VI-S, lyophilized powder, 200-1,000 units/mg protein
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.9 atom % D
Sigma-Aldrich
Butyrylcholinesterase from equine serum, lyophilized powder, ≥10 units/mg protein
Sigma-Aldrich
9-Amino-1,2,3,4-tetrahydroacridine hydrochloride hydrate, ≥99%
Sigma-Aldrich
5,5′-Dithiobis(2-nitrobenzoic acid), ≥98%, BioReagent, suitable for determination of sulfhydryl groups