- Adenovirus serotype 14 infection, New Brunswick, Canada, 2011.
Adenovirus serotype 14 infection, New Brunswick, Canada, 2011.
Emerging infectious diseases (2012-12-25)
Gabriel Girouard, Richard Garceau, Louise Thibault, Youcef Oussedik, Nathalie Bastien, Yan Li
PMID23260201
ABSTRACT
We describe 3 culture-proven cases of adenovirus serotype 14 infection in New Brunswick, Canada, during the summer of 2011. Strains isolated from severely ill patients were closely related to strains of a genomic variant, adenovirus 14p1, circulating in the United States and Ireland. Physicians in Canada should be aware of this emerging adenovirus.