Merck
  • New efficient substrates for semicarbazide-sensitive amine oxidase/VAP-1 enzyme: analysis by SARs and computational docking.
New efficient substrates for semicarbazide-sensitive amine oxidase/VAP-1 enzyme: analysis by SARs and computational docking.
Journal of medicinal chemistry (2006-10-13)
Francesc Yraola, Silvia García-Vicente, Juan Fernandez-Recio, Fernando Albericio, Antonio Zorzano, Luc Marti, Miriam Royo
ABSTRACT
Structure activity relationships for semicarbazide-sensitive amine oxidase/vascular adhesion protein-1 (SSAO/VAP-1) were studied using a library of arylalkylamine substrates, with the aim of contributing to the discovery of more efficient SSAO substrates. Experimental data were contrasted with computational docking studies, thereby allowing us to examine the mechanism and substrate-binding affinity of SSAO and thus contribute to the discovery of more efficient SSAO substrates and provide a structural basis for their interactions. We also built a model of the mouse SSAO structure, which provides several structural rationales for interspecies differences in SSAO substrate selectivity and reveals new trends in SSAO substrate recognition. In this context, we identified novel efficient substrates for human SSAO that can be used as a lead for the discovery of antidiabetic agents.
Sigma-Aldrich
Benzylamine, ReagentPlus®, 99%
Sigma-Aldrich
4-Methylbenzylamine, 97%
Sigma-Aldrich
m-Xylylenediamine, 99%
Sigma-Aldrich
4-Methoxybenzylamine, 98%
Sigma-Aldrich
4-(Aminomethyl)benzoic acid, 97%
Sigma-Aldrich
Phenethylamine, purified by redistillation, ≥99.5%
Sigma-Aldrich
4-tert-Butylbenzylamine, 97%
Sigma-Aldrich
3-Phenyl-1-propylamine, 98%
Sigma-Aldrich
Benzylamine, purified by redistillation, ≥99.5%
Supelco
Benzylamine, for GC derivatization, LiChropur, ≥99.0%
Sigma-Aldrich
Phenethylamine, ≥99%
Sigma-Aldrich
p-Xylylenediamine, 99%
Sigma-Aldrich
4-Fluorobenzylamine, 97%
Supelco
2-Phenethylamine, analytical standard