Merck
  • Generation of Human Renal Vesicles in Mouse Organ Niche Using Nephron Progenitor Cell Replacement System.
Generation of Human Renal Vesicles in Mouse Organ Niche Using Nephron Progenitor Cell Replacement System.
Cell reports (2020-09-17)
Toshinari Fujimoto, Shuichiro Yamanaka, Susumu Tajiri, Tsuyoshi Takamura, Yatsumu Saito, Naoto Matsumoto, Kei Matsumoto, Toshiaki Tachibana, Hirotaka James Okano, Takashi Yokoo
ABSTRACT
Animal fetuses may be used for the regeneration of human organs. We have previously generated a transgenic mouse model that allows diphtheria toxin (DT)-induced ablation of Six2-positive nephron progenitor cells (NPCs). Elimination of existing native host NPCs enables their replacement with donor NPCs, which can generate neo-nephrons. However, this system cannot be applied to human NPCs, because DT induces apoptosis in human cells. Therefore, the present study presents a transgenic mouse model for the ablation of NPCs using tamoxifen, which does not affect human cells. Using this system, we successfully regenerate interspecies neo-nephrons, which exhibit urine-producing abilities, from transplanted rat NPCs in a mouse host. Transplantation of human induced pluripotent stem cell (iPSC)-derived NPCs results in differentiation into renal vesicles, which connect to the ureteric bud of the host. Thus, we demonstrate the possibility of the regeneration of human kidneys derived from human iPSC-derived NPCs via NPC replacement.
Sigma-Aldrich
Anti-CDH6 antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium - high glucose, With 4500 mg/L glucose, L-glutamine, and sodium bicarbonate, without sodium pyruvate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Retinoic acid, ≥98% (HPLC), powder
Sigma-Aldrich
Anti-Aquaporin 1 Antibody, 1.0 mg/mL, Chemicon®
Sigma-Aldrich
Anti-Nuclei Antibody, clone 235-1, clone 235-1, Chemicon®, from mouse