Merck
  • Delayed Injection of a Physically Cross-Linked PNIPAAm-g-PEG Hydrogel in Rat Contused Spinal Cord Improves Functional Recovery.
Delayed Injection of a Physically Cross-Linked PNIPAAm-g-PEG Hydrogel in Rat Contused Spinal Cord Improves Functional Recovery.
ACS omega (2020-05-20)
Maxime Bonnet, Olivier Alluin, Thomas Trimaille, Didier Gigmes, Tanguy Marqueste, Patrick Decherchi
ABSTRACT
Spinal cord injury is a main health issue, leading to multiple functional deficits with major consequences such as motor and sensitive impairment below the lesion. To date, all repair strategies remain ineffective. In line with the experiments showing that implanted hydrogels, immunologically inert biomaterials, from natural or synthetic origins, are promising tools and in order to reduce functional deficits, to increase locomotor recovery, and to reduce spasticity, we injected into the lesion area, 1 week after a severe T10 spinal cord contusion, a thermoresponsive physically cross-linked poly(N-isopropylacrylamide)-poly(ethylene glycol) copolymer hydrogel. The effect of postinjury intensive rehabilitation training was also studied. A group of male Sprague-Dawley rats receiving the hydrogel was enrolled in an 8 week program of physical activity (15 min/day, 5 days/week) in order to verify if the combination of a treadmill step-training and hydrogel could lead to better outcomes. The data obtained were compared to those obtained in animals with a spinal lesion alone receiving a saline injection with or without performing the same program of physical activity. Furthermore, in order to verify the biocompatibility of our designed biomaterial, an inflammatory reaction (interleukin-1β, interleukin-6, and tumor necrosis factor-α) was examined 15 days post-hydrogel injection. Functional recovery (postural and locomotor activities and sensorimotor coordination) was assessed from the day of injection, once a week, for 9 weeks. Finally, 9 weeks postinjection, the spinal reflexivity (rate-dependent depression of the H-reflex) was measured. The results indicate that the hydrogel did not induce an additional inflammation. Furthermore, we observed the same significant locomotor improvements in hydrogel-injected animals as in trained saline-injected animals. However, the combination of hydrogel with exercise did not show higher recovery compared to that evaluated by the two strategies independently. Finally, the H-reflex depression recovery was found to be induced by the hydrogel and, albeit to a lesser degree, exercise. However, no recovery was observed when the two strategies were combined. Our results highlight the effectiveness of our copolymer and its high therapeutic potential to preserve/repair the spinal cord after lesion.
Product Description
Buprenorphine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Rat IL-6 ELISA Kit, for serum, plasma and cell culture supernatants
Sigma-Aldrich
Rat Tumor Necrosis Factor α ELISA Kit, for cell and tissue lysates
Sigma-Aldrich
Rat IL-1 α ELISA Kit, for serum, plasma and cell culture supernatant