Merck
  • Small molecule JQ1 promotes prostate cancer invasion via BET-independent inactivation of FOXA1.
Small molecule JQ1 promotes prostate cancer invasion via BET-independent inactivation of FOXA1.
The Journal of clinical investigation (2019-12-25)
Leiming Wang, Mafei Xu, Chung-Yang Kao, Sophia Y Tsai, Ming-Jer Tsai
ABSTRACT
Recent findings have shown that inhibitors targeting bromodomain and extraterminal domain (BET) proteins, such as the small molecule JQ1, are potent growth inhibitors of many cancers and hold promise for cancer therapy. However, some reports have also revealed that JQ1 can activate additional oncogenic pathways and may affect epithelial-to-mesenchymal transition (EMT). Therefore, it is important to address the potential unexpected effect of JQ1 treatment, such as cell invasion and metastasis. Here, we showed that in prostate cancer, JQ1 inhibited cancer cell growth but promoted invasion and metastasis in a BET protein-independent manner. Multiple invasion pathways including EMT, bone morphogenetic protein (BMP) signaling, chemokine signaling, and focal adhesion were activated by JQ1 to promote invasion. Notably, JQ1 induced upregulation of invasion genes through inhibition of Forkhead box protein A1 (FOXA1), an invasion suppressor in prostate cancer. JQ1 directly interacted with FOXA1 and inactivated FOXA1 binding to its interacting repressors TLE3, HDAC7, and NFIC, thereby blocking FOXA1-repressive function and activating the invasion genes. Our findings indicate that JQ1 has an unexpected effect of promoting invasion in prostate cancer. Thus, the ill effect of JQ1 or its derived therapeutic agents cannot be ignored during cancer treatment, especially in FOXA1-related cancers.
Sigma-Aldrich
Fetal Bovine Serum, USA origin, sterile-filtered, suitable for cell culture, suitable for hybridoma
Sigma-Aldrich
Fetal Bovine Serum, USA origin, Charcoal Stripped, sterile-filtered, suitable for cell culture
Sigma-Aldrich
GSK1210151A, ≥98% (HPLC)
Sigma-Aldrich
I-BET762, ≥98% (HPLC)
Sigma-Aldrich
(+)-Sodium L-ascorbate, BioXtra, ≥99.0% (NT)
Sigma-Aldrich
(+/-)-JQ1, ≥98% (HPLC)
Sigma-Aldrich
LDN-212854, ≥98% (HPLC)
Sigma-Aldrich
OTX015, ≥98% (HPLC)
Sigma-Aldrich
(-)-JQ1, ≥95% (HPLC)
Sigma-Aldrich
Azide-PEG3-biotin conjugate