Merck
  • RAP1-RAC1 Signaling Has an Important Role in Adhesion and Migration in HNSCC.
RAP1-RAC1 Signaling Has an Important Role in Adhesion and Migration in HNSCC.
Journal of dental research (2020-05-14)
M Liu, R Banerjee, C Rossa, N J D'Silva
ABSTRACT
Cell-cell adhesion is a key mechanism to control tissue integrity and migration. In head and neck squamous cell carcinoma (HNSCC), cell migration facilitates distant metastases and is correlated with poor prognosis. RAP1, a ras-like protein, has an important role in the progression of HNSCC. RAC1 is an integrin-linked, ras-like protein that promotes cell migration. Here we show that loss of cell-cell adhesion is correlated with inactivation of RAP1 confirmed by 2 different biochemical approaches. RAP1 activation is required for cell-matrix adhesion confirmed by adhesion to fibronectin-coated plates with cells that have biochemically activated RAP1. This effect is reversed when RAP1 is inactivated. In addition, RAP1GTP-mediated adhesion is only facilitated through α5β1 integrin complex and is not a function of either α5 or β1 integrin alone. Moreover, the inside-out signaling of RAP1 activation is coordinated with RAC1 activation. These findings show that RAP1 has a prominent role in cell-matrix adhesion via extracellular matrix molecule fibronectin-induced α5β1 integrin and supports a critical role for the RAP1/RAC1 signaling axis in HNSCC cell migration.
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, essentially fatty acid free and essentially globulin free, ≥99% (agarose gel electrophoresis)
Sigma-Aldrich
Anti-RPL23 antibody produced in rabbit, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-Integrin β1 Antibody, clone 6S6, Azide Free, clone 6S6, Chemicon®, from mouse
Sigma-Aldrich
Anti-Integrin α5 Antibody, clone P1D6, azide free, clone P1D6, Chemicon®, from mouse
Sigma-Aldrich
Anti-Rac1 Antibody, clone 23A8, clone 23A8, Upstate®, from mouse
Sigma-Aldrich
Anti-Glyceraldehyde-3-Phosphate Dehydrogenase Antibody, clone 6C5, clone 6C5, Chemicon®, from mouse