Merck
  • Anti-inflammatory effects of doxepin hydrochloride against LPS-induced C6-glioma cell inflammatory reaction by PI3K-mediated Akt signaling.
Anti-inflammatory effects of doxepin hydrochloride against LPS-induced C6-glioma cell inflammatory reaction by PI3K-mediated Akt signaling.
Journal of biochemical and molecular toxicology (2019-11-20)
Feng Tao, Jie Zhu, Lijie Duan, Junfeng Wu, Jianfeng Zhang, Kai Yao, Jimei Bo, Hengbing Zu
ABSTRACT
Recent studies have shown that tricyclic antidepressants (TCAs) may have anti-inflammatory and anticonvulsant effects in addition to its antidepressant effects. So far, the nonantidepressant effects of TCAs and their molecular pharmacological mechanisms remain completely unclear. Chronic inflammation in the brain parenchyma may be related to the pathogenesis and progression of various neurodegenerative diseases. As a common antidepressant and anti-insomnia drug, doxepin also may be a potential anti-inflammatory and anticonvulsant drug, so the study on the anti-inflammatory protective effect of doxepin and its molecular mechanism has become a very important issue in pharmacology and clinical medicine. Further elucidating the anti-inflammatory and neuroprotective effects of doxepin and its molecular mechanism may provide the important theoretical and clinical basis for the prevention and treatment of neurodegenerative disease. This study was designed to understand the glio-protective mechanism of doxepin against the inflammatory damage induced by lipopolysaccharide (LPS) exposure in C6-glioma cells. We found the treatment of C6-glioma cells with LPS results in deleterious effects, including the augmentation of inflammatory cytokine levels (tumor necrosis factor-α, interleukin-1β), and suppresses the Akt phosphorylation. Furthermore, our outcomes demonstrated that doxepin was able to suppress these effects induced by LPS, through activation of the phosphatidylinositol-3-kinase-mediated protein kinase B (Akt) pathway. To sum up, these results highlight the potential role of doxepin against neuroinflammatory-related disease in the brain.
Sigma-Aldrich
Octoclothepin maleate salt, solid
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O111:B4, purified by phenol extraction
Doxepin hydrochloride, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Anti-GAPDH antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution