Merck
  • The protective effect of PPARγ in sepsis-induced acute lung injury via inhibiting PTEN/β-catenin pathway.
The protective effect of PPARγ in sepsis-induced acute lung injury via inhibiting PTEN/β-catenin pathway.
Bioscience reports (2020-05-19)
Lili Liu, Junyi Chen, Xiaofang Zhang, Xue Cui, Nana Qiao, Yun Zhang, Jie Yang
ABSTRACT
The present study aims to reveal the molecular mechanism of peroxisome proliferator-activated receptor γ (PPARγ) on sepsis-induced acute lung injury (ALI). To do that, the rat injury model was established using cecal ligation and perforation (CLP) method, followed by different treatments, and the rats were divided into Sham group, CLP group, CLP + rosiglitazone (PPARγ agonist) group, CLP + GW9662 (PPARγ inhibitor) group, CLP + bpV (phosphatase and tensin homolog (PTEN) inhibitor) group, CLP + GW9662 + bpV group. Compared with Sham group, the mRNA and protein expression levels of PPARγ were down-regulated, the inflammation levels were elevated, and the apoptosis was increased in CLP group. After treatment with rosiglitazone, the protein expression level of PPARγ was significantly up-regulated, the phosphorylation level of PTEN/β-catenin pathway was decreased, the PTEN/β-catenin pathway was inhibited, the lung injury, inflammation and apoptosis were reduced. The opposite effect was observed after treatment with GW9662. Besides, bpV inhibited PTEN/β-catenin pathway, and relieved the lung tissue injury. The overexpression of PPARγ reduced inflammatory response and inhibited apoptosis in sepsis-induced ALI. Furthermore, PPARγ relieved the sepsis-induced ALI by inhibiting the PTEN/β-catenin pathway.
Sigma-Aldrich
Monoclonal Anti-β-Catenin antibody produced in mouse, clone 6F9, ascites fluid
Sigma-Aldrich
Anti-PPAR γ Antibody, isoform 1&2, ascites fluid, clone 1H4, Chemicon®
Sigma-Aldrich
Anti-AKT antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-phospho-Akt (pSer124) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Rosiglitazone, ≥98% (HPLC)
Sigma-Aldrich
Anti-PTEN (Ab-370) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-phospho-PTEN (pSer380/pThr382/pThr383) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-phospho-Glycogen Synthase Kinase 3α/β (pTyr279/216) antibody produced in rabbit, affinity isolated antibody, aqueous glycerol solution
Sigma-Aldrich
Anti-Glycogen Synthase Kinase-3β (GSK-3β) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution