Merck
  • ATF4-Dependent NRF2 Transcriptional Regulation Promotes Antioxidant Protection during Endoplasmic Reticulum Stress.
ATF4-Dependent NRF2 Transcriptional Regulation Promotes Antioxidant Protection during Endoplasmic Reticulum Stress.
Cancers (2020-03-04)
Carmen Sarcinelli, Helena Dragic, Marie Piecyk, Virginie Barbet, Cédric Duret, Audrey Barthelaix, Carole Ferraro-Peyret, Joelle Fauvre, Toufic Renno, Cédric Chaveroux, Serge N Manié
ABSTRACT
Endoplasmic reticulum (ER) stress generates reactive oxygen species (ROS) that induce apoptosis if left unabated. To limit oxidative insults, the ER stress PKR-like endoplasmic reticulum Kinase (PERK) has been reported to phosphorylate and activate nuclear factor erythroid 2-related factor 2 (NRF2). Here, we uncover an alternative mechanism for PERK-mediated NRF2 regulation in human cells that does not require direct phosphorylation. We show that the activation of the PERK pathway rapidly stimulates the expression of NRF2 through activating transcription factor 4 (ATF4). In addition, NRF2 activation is late and largely driven by reactive oxygen species (ROS) generated during late protein synthesis recovery, contributing to protecting against cell death. Thus, PERK-mediated NRF2 activation encompasses a PERK-ATF4-dependent control of NRF2 expression that contributes to the NRF2 protective response engaged during ER stress-induced ROS production.
Product Description
SAFC
DMEM/Nutrient Mixture F-12 Ham, With 15 mM HEPES, without L-glutamine, L-leucine, L-lysine, L-methionine, CaCl2, MgCl2, MgSO4, sodium bicarbonate, and phenol red, powder, suitable for cell culture
Sigma-Aldrich
Thapsigargin, ≥98% (HPLC), solid film
Sigma-Aldrich
Puromycin dihydrochloride, Ready Made Solution, from Streptomyces alboniger, 10 mg/mL in H2O, suitable for cell culture
Sigma-Aldrich
Anti-Puromycin Antibody, clone 12D10, clone 12D10, from mouse
Supelco
Hydrogen peroxide 30%, (Perhydrol) for analysis EMSURE ISO
Sigma-Aldrich
MISSION® esiRNA, targeting human ATF4
Sigma-Aldrich
Tunicamycin from Streptomyces sp.