Merck
  • Molecular Discrimination between Two Conformations of Sphingomyelin in Plasma Membranes.
Molecular Discrimination between Two Conformations of Sphingomyelin in Plasma Membranes.
Cell (2019-02-05)
Shreya Endapally, Donna Frias, Magdalena Grzemska, Austin Gay, Diana R Tomchick, Arun Radhakrishnan
ABSTRACT
Sphingomyelin and cholesterol are essential lipids that are enriched in plasma membranes of animal cells, where they interact to regulate membrane properties and many intracellular signaling processes. Despite intense study, the interaction between these lipids in membranes is not well understood. Here, structural and biochemical analyses of ostreolysin A (OlyA), a protein that binds to membranes only when they contain both sphingomyelin and cholesterol, reveal that sphingomyelin adopts two distinct conformations in membranes when cholesterol is present. One conformation, bound by OlyA, is induced by stoichiometric, exothermic interactions with cholesterol, properties that are consistent with sphingomyelin/cholesterol complexes. In its second conformation, sphingomyelin is free from cholesterol and does not bind OlyA. A point mutation abolishes OlyA's ability to discriminate between these two conformations. In cells, levels of sphingomyelin/cholesterol complexes are held constant over a wide range of plasma membrane cholesterol concentrations, enabling precise regulation of the chemical activity of cholesterol.
Product Description
Avanti
18:1 SM (d18:1/18:1(9Z)), Avanti Research - A Croda Brand
Avanti
W140, (S)-3-amino-4-(3-hexylphenylamino)-4-oxobutylphosphonic acid (TFA salt), powder
Avanti
16:0 SM (d18:1/16:0), Avanti Research - A Croda Brand, powder
Avanti
06:0 SM (d18:1/6:0), Avanti Research - A Croda Brand 860582C
Avanti
18:0 SM (d18:1/18:0), Avanti Research - A Croda Brand
Avanti
06:0 SM (d18:1/6:0), Avanti Research - A Croda Brand 860582P, powder
Avanti
Ganglioside-Total, Avanti Research - A Croda Brand
Sigma-Aldrich
Anti-Histidine Tagged Antibody, clone HIS.H8, clone HIS.H8, Upstate®, from mouse