Merck
  • Adipocyte Hypoxia-Inducible Factor 2α Suppresses Atherosclerosis by Promoting Adipose Ceramide Catabolism.
Adipocyte Hypoxia-Inducible Factor 2α Suppresses Atherosclerosis by Promoting Adipose Ceramide Catabolism.
Cell metabolism (2019-11-02)
Xingzhong Zhang, Yangming Zhang, Pengcheng Wang, Song-Yang Zhang, Yongqiang Dong, Guangyi Zeng, Yu Yan, Lulu Sun, Qing Wu, Huiying Liu, Bo Liu, Wei Kong, Xian Wang, Changtao Jiang
ABSTRACT
Obesity-induced adipose dysfunction is a major contributor to atherosclerosis. Cold exposure has been reported to affect atherosclerosis through regulation of adipose function, but the mechanism has not been well clarified. Here, adipocyte hypoxia-inducible factor 2α (HIF-2α) was upregulated after mild cold exposure at 16°C and mediated cold-induced thermogenesis. Adipocyte HIF-2α deficiency exacerbated Western-diet-induced atherosclerosis by increasing adipose ceramide levels, which blunted hepatocyte cholesterol elimination and thermogenesis. Mechanistically, Acer2, the gene encoding alkaline ceramidase 2, was identified as a novel target gene of HIF-2α, triggering ceramide catabolism. Adipose overexpression of ACER2 rescued adipocyte HIF-2α-deficiency-induced exacerbation of atherosclerosis. Furthermore, activation of adipose HIF-2α by the HIF prolyl hydroxylase inhibitor FG-4592 had protective effects on atherosclerosis, accompanied by a reduction in adipose and plasma ceramide and plasma cholesterol levels. This study highlights adipocyte HIF-2α as a putative drug target against atherosclerosis.
Product Description
Avanti
C18 Dihydroceramide (d18:0/18:0), Avanti Research - A Croda Brand 860627P, powder
Avanti
C20 Ceramide (d18:1/20:0), Avanti Research - A Croda Brand
Avanti
18:0 SM (d18:1/18:0), Avanti Research - A Croda Brand
Avanti
C24:1 Dihydroceramide (d18:0/24:1(15Z)), Avanti Research - A Croda Brand 860629P, powder
Avanti
C14 dihydroceramide (d18:0/14:0), Avanti Research - A Croda Brand 860632P, powder
Avanti
16:0 SM (d18:1/16:0), Avanti Research - A Croda Brand, powder
Avanti
C22 Ceramide (d18:1/22:0), Avanti Research - A Croda Brand 860501P, powder