Merck
  • Selective organ targeting (SORT) nanoparticles for tissue-specific mRNA delivery and CRISPR-Cas gene editing.
Selective organ targeting (SORT) nanoparticles for tissue-specific mRNA delivery and CRISPR-Cas gene editing.
Nature nanotechnology (2020-04-07)
Qiang Cheng, Tuo Wei, Lukas Farbiak, Lindsay T Johnson, Sean A Dilliard, Daniel J Siegwart
ABSTRACT
CRISPR-Cas gene editing and messenger RNA-based protein replacement therapy hold tremendous potential to effectively treat disease-causing mutations with diverse cellular origin. However, it is currently impossible to rationally design nanoparticles that selectively target specific tissues. Here, we report a strategy termed selective organ targeting (SORT) wherein multiple classes of lipid nanoparticles are systematically engineered to exclusively edit extrahepatic tissues via addition of a supplemental SORT molecule. Lung-, spleen- and liver-targeted SORT lipid nanoparticles were designed to selectively edit therapeutically relevant cell types including epithelial cells, endothelial cells, B cells, T cells and hepatocytes. SORT is compatible with multiple gene editing techniques, including mRNA, Cas9 mRNA/single guide RNA and Cas9 ribonucleoprotein complexes, and is envisioned to aid the development of protein replacement and gene correction therapeutics in targeted tissues.
Product Description
Avanti
18:1 (Δ9-Cis) PC (DOPC), Avanti Research - A Croda Brand
Avanti
18:1 (Δ9-Cis) PC (DOPC), Avanti Research - A Croda Brand
Avanti
14:0 PA, 1,2-dimyristoyl-sn-glycero-3-phosphate (sodium salt), powder
Avanti
18:1 TAP (DOTAP), Avanti Research - A Croda Brand
Avanti
18:0 DDAB, Avanti Research - A Croda Brand 890810C
Avanti
18:0 DDAB, Avanti Research - A Croda Brand
Avanti
18:1 Hemi BMP (S,R), sn-(3-oleoyl-2-hydroxy)-glycerol-1-phospho-sn-3′-(1′,2′-dioleoyl)-glycerol (ammonium salt), powder
Avanti
18:1 TAP (DOTAP), Avanti Research - A Croda Brand
Sigma-Aldrich
L-α-Phosphatidylethanolamine, dioleoyl, ≥99% (GC), ≥98% (TLC), lyophilized powder
Avanti
Egg PC, Avanti Research - A Croda Brand
Sigma-Aldrich
Didecyldimethylammonium bromide, 98%
Avanti
18:1 DAP, Avanti Research - A Croda Brand
Avanti
18:0 PA, Avanti Research - A Croda Brand
Avanti
Egg PC, Avanti Research - A Croda Brand
Avanti
18:1 DAP, Avanti Research - A Croda Brand 890850C
Avanti
18:1 (Δ9-Cis) PE (DOPE), Avanti Research - A Croda Brand
Avanti
18:1 (Δ9-Cis) PE (DOPE), Avanti Research - A Croda Brand
Avanti
18:0 PC (DSPC), Avanti Research - A Croda Brand
Avanti
18:0 PC (DSPC), Avanti Research - A Croda Brand