Merck
  • Cascade synthesis of rare ketoses by whole cells based on L-rhamnulose-1-phosphate aldolase.
Cascade synthesis of rare ketoses by whole cells based on L-rhamnulose-1-phosphate aldolase.
Enzyme and microbial technology (2019-12-26)
Zhou Chen, Zijie Li, Fen Li, Mayan Wang, Ning Wang, Xiao-Dong Gao
ABSTRACT
Dihydroxyacetone phosphate (DHAP)-dependent aldolases demonstrate important values in the production of rare ketoses due to their unique stereoselectivities. As a specific example, we developed an efficient Escherichia coli whole-cell biocatalytic cascade system in which rare ketoses were produced from abundant glycerol and catalyzed by four enzymes based on L-rhamnulose-1-phosphate aldolase (RhaD). For the semicontinuous bioconversion in which D-glyceraldehyde was continuously added, once D-glyceraldehyde was consumed, the final yields of D-sorbose and D-psicose were 15.30 g/L and 6.35 g/L, respectively. Moreover, the maximum conversion rate and productivity of D-sorbose and D-psicose were 99% and 1.11 g/L/h at 8 h, respectively. When L-glyceraldehyde was used instead of the D-isomer, the final yield of L-fructose was 16.80 g/L. Furthermore, the maximum conversion rate and productivity of L-fructose were 95% and 1.08 g/L/h at 8 h, respectively. This synthetic platform was also compatible with other various aldehydes, which allowed the production of many other high-value chemicals from glycerol.
Sigma-Aldrich
D-(+)-Sorbose, ≥99%
Sigma-Aldrich
L-(+)-Fructose, ≥97.0% (HPLC)
Sigma-Aldrich
D-Psicose, ≥95% (HPLC)
Sigma-Aldrich
L-(−)-Glyceraldehyde, ≥90% (HPLC)
Sigma-Aldrich
D-(+)-Glyceraldehyde, ≥98.0% (HPLC)