Merck
  • The Repertoire of Serous Ovarian Cancer Non-genetic Heterogeneity Revealed by Single-Cell Sequencing of Normal Fallopian Tube Epithelial Cells.
The Repertoire of Serous Ovarian Cancer Non-genetic Heterogeneity Revealed by Single-Cell Sequencing of Normal Fallopian Tube Epithelial Cells.
Cancer cell (2020-02-13)
Zhiyuan Hu, Mara Artibani, Abdulkhaliq Alsaadi, Nina Wietek, Matteo Morotti, Tingyan Shi, Zhe Zhong, Laura Santana Gonzalez, Salma El-Sahhar, Mohammad KaramiNejadRanjbar, Garry Mallett, Yun Feng, Kenta Masuda, Yiyan Zheng, Kay Chong, Stephen Damato, Sunanda Dhar, Leticia Campo, Riccardo Garruto Campanile, Hooman Soleymani Majd, Vikram Rai, David Maldonado-Perez, Stephanie Jones, Vincenzo Cerundolo, Tatjana Sauka-Spengler, Christopher Yau, Ahmed Ashour Ahmed
ABSTRACT
The inter-differentiation between cell states promotes cancer cell survival under stress and fosters non-genetic heterogeneity (NGH). NGH is, therefore, a surrogate of tumor resilience but its quantification is confounded by genetic heterogeneity. Here we show that NGH in serous ovarian cancer (SOC) can be accurately measured when informed by the molecular signatures of the normal fallopian tube epithelium (FTE) cells, the cells of origin of SOC. Surveying the transcriptomes of ∼6,000 FTE cells, predominantly from non-ovarian cancer patients, identified 6 FTE subtypes. We used subtype signatures to deconvolute SOC expression data and found substantial intra-tumor NGH. Importantly, NGH-based stratification of ∼1,700 tumors robustly correlated with survival. Our findings lay the foundation for accurate prognostic and therapeutic stratification of SOC.
Sigma-Aldrich
Anti-SPARC antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-CCDC17 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-CAPS antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-KRT17 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-RGS16 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Monoclonal Anti-β-Tubulin IV antibody produced in mouse, clone ONS.1A6, ascites fluid
Sigma-Aldrich
Anti-Cytokeratin 7 Antibody, clone LP5K, FITC conjugated, clone LP5K, Chemicon®, from mouse