Merck
  • Ultrastructure of light-activated axons following optogenetic stimulation to produce late-phase long-term potentiation.
Ultrastructure of light-activated axons following optogenetic stimulation to produce late-phase long-term potentiation.
PloS one (2020-01-16)
Masaaki Kuwajima, Olga I Ostrovskaya, Guan Cao, Seth A Weisberg, Kristen M Harris, Boris V Zemelman
ABSTRACT
Analysis of neuronal compartments has revealed many state-dependent changes in geometry but establishing synapse-specific mechanisms at the nanoscale has proven elusive. We co-expressed channelrhodopsin2-GFP and mAPEX2 in a subset of hippocampal CA3 neurons and used trains of light to induce late-phase long-term potentiation (L-LTP) in area CA1. L-LTP was shown to be specific to the labeled axons by severing CA3 inputs, which prevented back-propagating recruitment of unlabeled axons. Membrane-associated mAPEX2 tolerated microwave-enhanced chemical fixation and drove tyramide signal amplification to deposit Alexa Fluor dyes in the light-activated axons. Subsequent post-embedding immunogold labeling resulted in outstanding ultrastructure and clear distinctions between labeled (activated), and unlabeled axons without obscuring subcellular organelles. The gold-labeled axons in potentiated slices were reconstructed through serial section electron microscopy; presynaptic vesicles and other constituents could be quantified unambiguously. The genetic specification, reliable physiology, and compatibility with established methods for ultrastructural preservation make this an ideal approach to link synapse ultrastructure and function in intact circuits.
Sigma-Aldrich
Anti-c-Myc antibody produced in rabbit, ~0.5 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
p-Xylene-bis(N-pyridinium bromide), ≥95% (TLC)
Sigma-Aldrich
DNQX, ≥98% (TLC)
Sigma-Aldrich
Anti-Cy5 antibody, Mouse monoclonal, clone CY5-15, purified from hybridoma cell culture
Sigma-Aldrich
Triton X-100, laboratory grade