Merck
  • Requirement for p62 acetylation in the aggregation of ubiquitylated proteins under nutrient stress.
Requirement for p62 acetylation in the aggregation of ubiquitylated proteins under nutrient stress.
Nature communications (2019-12-21)
Zhiyuan You, Wen-Xue Jiang, Ling-Yun Qin, Zhou Gong, Wei Wan, Jin Li, Yusha Wang, Hongtao Zhang, Chao Peng, Tianhua Zhou, Chun Tang, Wei Liu
ABSTRACT
Autophagy receptor p62/SQSTM1 promotes the assembly and removal of ubiquitylated proteins by forming p62 bodies and mediating their encapsulation in autophagosomes. Here we show that under nutrient-deficient conditions, cellular p62 specifically undergoes acetylation, which is required for the formation and subsequent autophagic clearance of p62 bodies. We identify K420 and K435 in the UBA domain as the main acetylation sites, and TIP60 and HDAC6 as the acetyltransferase and deacetylase. Mechanically, acetylation at both K420 and K435 sites enhances p62 binding to ubiquitin by disrupting UBA dimerization, while K435 acetylation also directly increases the UBA-ubiquitin affinity. Furthermore, we show that acetylation of p62 facilitates polyubiquitin chain-induced p62 phase separation. Our results suggest an essential role of p62 acetylation in the selective degradation of ubiquitylated proteins in cells under nutrient stress, by specifically regulating the assembly of p62 bodies.
Sigma-Aldrich
Anti-ATG7 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
O-Phospho-L-tyrosine
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, ascites fluid
Sigma-Aldrich
Thrombin from bovine plasma, lyophilized powder, 40-300 NIH units/mg protein (biuret)
Sigma-Aldrich
Saponin from quillaja bark, Sapogenin content ≥10 %
Sigma-Aldrich
Nitrilotriacetic acid disodium salt, Sigma Grade, ≥99%
Sigma-Aldrich
Monoclonal Anti-β-Tubulin antibody produced in mouse, clone 2-28-33, ascites fluid
Sigma-Aldrich
Ammonium-15N chloride, ≥98 atom % 15N, ≥99% (CP)
Sigma-Aldrich
Anti-Acetyl-alpha-Tubulin (Lys40) antibody, Rabbit monoclonal, recombinant, expressed in HEK 293 cells, clone RM318, purified immunoglobulin
SAFC
Isopropyl β-D-1-thiogalactopyranoside