Merck
  • Serotonin 5-HT2C receptor protein expression is enriched in synaptosomal and post-synaptic compartments of rat cortex.
Serotonin 5-HT2C receptor protein expression is enriched in synaptosomal and post-synaptic compartments of rat cortex.
Journal of neurochemistry (2010-03-30)
Noelle C Anastasio, Maria Fe Lanfranco, Marcy J Bubar, Patricia K Seitz, Sonja J Stutz, Andrew G McGinnis, Cheryl S Watson, Kathryn A Cunningham
ABSTRACT
The action of serotonin (5-HT) at the 5-HT(2C) receptor (5-HT(2C)R) in cerebral cortex is emerging as a candidate modulator of neural processes that mediate core phenotypic facets of several psychiatric and neurological disorders. However, our understanding of the neurobiology of the cortical 5-HT(2C)R protein complex is currently limited. The goal of the present study was to explore the subcellular localization of the 5-HT(2C)R in synaptosomes and the post-synaptic density, an electron-dense thickening specialized for post-synaptic signaling and neuronal plasticity. Utilizing multiples tissues (brain, peripheral tissues), protein fractions (synaptosomal, post-synaptic density), and controls (peptide neutralization, 5-HT(2C)R stably-expressing cells), we established the selectivity of two commercially available 5-HT(2C)R antibodies and employed the antibodies in western blot and immunoprecipitation studies of prefrontal cortex (PFC) and motor cortex, two regions implicated in cognitive, emotional and motor dysfunction. For the first time, we demonstrated the expression of the 5-HT(2C)R in post-synaptic density-enriched fractions from both PFC and motor cortex. Co-immunoprecipitation studies revealed the presence of post-synaptic density-95 within the 5-HT(2C)R protein complex expressed in PFC and motor cortex. Taken together, these data support the hypothesis that the 5-HT(2C)R is localized within the post-synaptic thickening of synapses and is therefore positioned to directly modulate synaptic plasticity in cortical neurons.
Sigma-Aldrich
Anti-Actin Antibody, clone C4, ascites fluid, clone C4, Chemicon®
Sigma-Aldrich
Anti-Post Synaptic Density Protein 95 Antibody, clone 7E3-1B8, clone 7E3-1B8, Chemicon®, from mouse
Sigma-Aldrich
Anti-SNAP-25 Antibody, Chemicon®, from goat
Sigma-Aldrich
Anti-Syntaxin Antibody, clone SP8, ascites fluid, clone SP8, Chemicon®