Merck
  • Physical inactivity affects skeletal muscle insulin signaling in a birth weight-dependent manner.
Physical inactivity affects skeletal muscle insulin signaling in a birth weight-dependent manner.
Journal of diabetes and its complications (2013-10-15)
Brynjulf Mortensen, Martin Friedrichsen, Nicoline R Andersen, Amra C Alibegovic, Lise Højbjerre, Mette P Sonne, Bente Stallknecht, Flemming Dela, Jørgen F P Wojtaszewski, Allan Vaag
ABSTRACT
We investigated whether physical inactivity could unmask defects in insulin and AMPK signaling in low birth weight (LBW) subjects. Twenty LBW and 20 normal birth weight (NBW) subjects were investigated using the euglycemic-hyperinsulinemic clamp with excision of skeletal muscle biopsies pre and post 9days of bed rest. Employing Western blotting, we investigated skeletal muscle Akt, AS160, GLUT4, and AMPK signaling. Peripheral insulin action was similar in the two groups and was decreased to the same extent post bed rest. Insulin and AMPK signaling was unaffected by bed rest in NBW individuals. LBW subjects showed decreased insulin-stimulated Akt phosphorylation and increased AMPK α1 and γ3 protein expression post bed rest. Insulin response of AS160 phosphorylation was lower in LBW subjects both pre and post bed rest. Bed rest-induced insulin resistance is not explained by impaired muscle insulin or AMPK signaling in subjects with or without LBW. Lower muscle insulin signaling in LBW subjects post bed rest despite similar degree of insulin resistance as seen in controls may to some extent support the idea that LBW subjects are at higher risk of developing type 2 diabetes when being physically inactive.
Sigma-Aldrich
Anti-Akt1/PKBα Antibody, Upstate®, from rabbit
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Anti-phospho-Acetyl CoA Carboxylase (Ser79) Antibody, Upstate®, from rabbit