Merck
  • Erbin and the NF2 tumor suppressor Merlin cooperatively regulate cell-type-specific activation of PAK2 by TGF-beta.
Erbin and the NF2 tumor suppressor Merlin cooperatively regulate cell-type-specific activation of PAK2 by TGF-beta.
Developmental cell (2009-03-18)
Mark C Wilkes, Claire E Repellin, Min Hong, Margarita Bracamonte, Sumedha G Penheiter, Jean-Paul Borg, Edward B Leof
ABSTRACT
Transforming growth factor beta (TGF-beta) family ligands are pleotropic proteins with diverse cell-type-specific effects on growth and differentiation. For example, PAK2 activation is critical for the proliferative/profibrotic action of TGF-beta on mesenchymal cells, and yet it is not responsive to TGF-beta in epithelial cells. We therefore investigated the regulatory constraints that prevent inappropriate PAK2 activation in epithelial cultures. The results show that the epithelial-enriched protein Erbin controls the function of the NF2 tumor suppressor Merlin by determining the output of Merlin's physical interactions with active PAK2. Whereas mesenchymal TGF-beta signaling induces PAK2-mediated inhibition of Merlin function in the absence of Erbin, Erbin/Merlin complexes bind and inactivate GTPase-bound PAK2 in epithelia. These results not only identify Erbin as a key determinant of epithelial resistance to TGF-beta signaling, they also show that Erbin controls Merlin tumor suppressor function by switching the functional valence of PAK2 binding.
Sigma-Aldrich
Anti-Glyceraldehyde-3-Phosphate Dehydrogenase Antibody, clone 6C5, clone 6C5, ChemiconĀ®, from mouse
Sigma-Aldrich
Cdc42 Activation Assay Kit, Non-radioactive cdc42 Activation Assay Kit can be used to precipitate cdc42-GTP from cell lysates subjected to various treatments & detection by a cdc42 specific monoclonal antibody.
Sigma-Aldrich
Anti-Rac1 Antibody, clone 23A8, clone 23A8, UpstateĀ®, from mouse