Merck
  • Cofactor molecules maintain infectious conformation and restrict strain properties in purified prions.
Cofactor molecules maintain infectious conformation and restrict strain properties in purified prions.
Proceedings of the National Academy of Sciences of the United States of America (2012-06-20)
Nathan R Deleault, Daniel J Walsh, Justin R Piro, Fei Wang, Xinhe Wang, Jiyan Ma, Judy R Rees, Surachai Supattapone
ABSTRACT
Prions containing misfolded prion protein (PrP(Sc)) can be formed with cofactor molecules using the technique of serial protein misfolding cyclic amplification. However, it remains unknown whether cofactors materially participate in maintaining prion conformation and infectious properties. Here we show that withdrawal of cofactor molecules during serial propagation of purified recombinant prions caused adaptation of PrP(Sc) structure accompanied by a reduction in specific infectivity of >10(5)-fold, to undetectable levels, despite the ability of adapted "protein-only" PrP(Sc) molecules to self-propagate in vitro. We also report that changing only the cofactor component of a minimal reaction substrate mixture during serial propagation induced major changes in the strain properties of an infectious recombinant prion. Moreover, propagation with only one functional cofactor (phosphatidylethanolamine) induced the conversion of three distinct strains into a single strain with unique infectious properties and PrP(Sc) structure. Taken together, these results indicate that cofactor molecules can regulate the defining features of mammalian prions: PrP(Sc) conformation, infectivity, and strain properties. These findings suggest that cofactor molecules likely are integral components of infectious prions.
Product Description
Avanti
C18(Plasm)-18:1 PE, 1-(1Z-octadecenyl)-2-oleoyl-sn-glycero-3-phosphoethanolamine, powder
Avanti
C18(Plasm)-18:1 PE, Avanti Research - A Croda Brand