Merck
Lipopolysaccharide induced conversion of recombinant prion protein.
Prion (2014-05-14)
Fozia Saleem, Trent C Bjorndahl, Carol L Ladner, Rolando Perez-Pineiro, Burim N Ametaj, David S Wishart
ABSTRACT
The conformational conversion of the cellular prion protein (PrP(C)) to the β-rich infectious isoform PrP(Sc) is considered a critical and central feature in prion pathology. Although PrP(Sc) is the critical component of the infectious agent, as proposed in the "protein-only" prion hypothesis, cellular components have been identified as important cofactors in triggering and enhancing the conversion of PrP(C) to proteinase K resistant PrP(Sc). A number of in vitro systems using various chemical and/or physical agents such as guanidine hydrochloride, urea, SDS, high temperature, and low pH, have been developed that cause PrP(C) conversion, their amplification, and amyloid fibril formation often under non-physiological conditions. In our ongoing efforts to look for endogenous and exogenous chemical mediators that might initiate, influence, or result in the natural conversion of PrP(C) to PrP(Sc), we discovered that lipopolysaccharide (LPS), a component of gram-negative bacterial membranes interacts with recombinant prion proteins and induces conversion to an isoform richer in β sheet at near physiological conditions as long as the LPS concentration remains above the critical micelle concentration (CMC). More significant was the LPS mediated conversion that was observed even at sub-molar ratios of LPS to recombinant ShPrP (90-232).
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O111:B4, purified by gel-filtration chromatography
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O111:B4, purified by ion-exchange chromatography, TLR ligand tested
Avanti
16:0-18:1 PG, Avanti Research - A Croda Brand
Avanti
16:0-18:1 PG, Avanti Research - A Croda Brand
Avanti
C18(Plasm)-18:1 PE, 1-(1Z-octadecenyl)-2-oleoyl-sn-glycero-3-phosphoethanolamine, powder
Avanti
C18(Plasm)-18:1 PE, Avanti Research - A Croda Brand