Merck
  • Rose Bengal Immobilized on Supported Ionic-Liquid-like Phases: An Efficient Photocatalyst for Batch and Flow Processes.
Rose Bengal Immobilized on Supported Ionic-Liquid-like Phases: An Efficient Photocatalyst for Batch and Flow Processes.
ChemSusChem (2019-07-10)
David Valverde, Raul Porcar, Diana Izquierdo, M Isabel Burguete, Eduardo Garcia-Verdugo, Santiago V Luis
ABSTRACT
The catalytic activity of Rose Bengal (RB) immobilized on supported ionic liquid (IL)-like phases was evaluated as a polymer-supported photocatalyst. In these systems, the polymer was designed to play a pivotal role. The polymeric backbone adequately modified with IL-like moieties (supported IL-like phases, SILLPs) was not just an inert support for the dye but controlled the accessibility of reagents/substrates to the active sites and provided specific microenvironments for the reaction. The structure of SILLPs could be finetuned to adjust the catalytic efficiency of the RB-SILLP composites, achieving systems that were more active and stable than the related systems in the absence of IL-like units.
Sigma-Aldrich
1-Decyl-2-methylimidazole, 97%
Sigma-Aldrich
N,N′-Methylenebis(acrylamide), 99%
Sigma-Aldrich
2,2′-Azobis(2-methylpropionitrile) solution, 0.2 M in toluene
Sigma-Aldrich
1-Butylimidazole, 98%
Sigma-Aldrich
Trimethylolpropane trimethacrylate, contains 250 ppm monomethyl ether hydroquinone as inhibitor, technical grade