Merck
  • The Duration of Nerve Block from Local Anesthetic Formulations in Male and Female Rats.
The Duration of Nerve Block from Local Anesthetic Formulations in Male and Female Rats.
Pharmaceutical research (2019-11-11)
Kathleen Cullion, Laura C Petishnok, Tianjiao Ji, David Zurakowski, Daniel S Kohane
ABSTRACT
It is unknown whether there are sex differences in response to free or encapsulated local anesthetics. We examined nerve block duration and toxicity following peripheral nerve blockade in male and female rats. We studied the local anesthetic bupivacaine (free or encapsulated) as well as tetrodotoxin, which acts on a different site of the same voltage-gated channel. Sensory nerve blockade was 158.5 [139-190] minutes (median [interquartile range]) (males) compared to 173 [134-171] minutes (females) (p = 0.702) following bupivacaine injection, N = 8 male, 8 female. Motor nerve blockade was 157 [141-171] minutes (males) compared to 172 [146-320] minutes (females) (p = 0.2786). Micellar bupivacaine (N = 8 male, 8 female) resulted in sensory nerve blockade of 266 [227-320] minutes (males) compared to 285 [239-344] minutes (females) (p = 0.6427). Motor nerve blockade was 264 [251-264] minutes (males) compared to 287 [262-287] minutes (females) (p = 0.3823). Liposomal bupivacaine (N = 8 male, 8 female) resulted in sensory nerve blockade of 240 [207-277] minutes (males) compared to 289 [204-348] minutes (females) (p = 0.1654). Motor nerve blockade was 266 [237-372] minutes (males) compared to 317 [251-356] minutes (females) (p = 0.6671). Following tetrodotoxin injection (N = 12 male,12 female) sensory nerve blockade was 54.8 [5-117] minutes (males) compared to 54 [14-71] minutes (females) (p = 0.6422). Motor nerve blockade was 72 [40-112] minutes (males) compared to 64 [32-143] minutes (females) (p = 0.971). We found no statistically significant sex differences associated with the formulations tested. In both sexes, durations of nerve block were similar between micellar and liposomal bupivacaine formulations, despite the micellar formulation containing less drug.
Product Description
Avanti
18:0 PEG2000 PE, Avanti Research - A Croda Brand 880120C
SKU
Pack Size
Availability
Price
Quantity
Avanti
18:0 PEG2000 PE, Avanti Research - A Croda Brand