Merck
  • Evaluation of Targeted Delivery to the Brain Using Magnetic Immunoliposomes and Magnetic Force.
Evaluation of Targeted Delivery to the Brain Using Magnetic Immunoliposomes and Magnetic Force.
Materials (Basel, Switzerland) (2019-11-07)
Louiza Bohn Thomsen, Thomas Linemann, Svend Birkelund, Gitte Abildgaard Tarp, Torben Moos
ABSTRACT
Magnetic nanoparticles have great prospects for drug delivery purposes, as they can be designed with various surface coatings and conjugated with drugs and targeting moieties. They also have a unique potential for precise delivery when guided by magnetic force. The blood-brain barrier (BBB) denotes the interface between the blood and brain parenchyma and hinders the majority of drugs from entering the brain. Red fluorescent magnetic nanoparticles were encapsulated in liposomes and conjugated to antibodies targeting the rat transferrin receptor (OX26) to form magnetic immunoliposomes. These magnetic immunoliposomes enhanced the uptake by rat brain capillary endothelial cells (BCECs) in vitro. In situ brain perfusion in young rats high in the endogenous expression of transferrin receptors by BCECs, revealed enhanced uptake of magnetic immunoliposomes when compared to naked magnetic nanoparticles or non-targeted magnetic liposomes. When applying the external magnetic force, the magnetic nanoparticles were detected in the brain parenchyma, suggesting transport across the BBB. Ultrastructural examination of the immunoliposomes, unfortunately, was unable to confirm a complete encapsulation of all naked nanoparticles within the liposomes, suggesting that the data on the brain could derive from particles being released from the liposomes under influence of external magnetic force; hence hypothesizes on external magnetic force as a qualifier for dragging targeted magnetic immunoliposomes through the BBB. In conclusion, our results suggest that transport of magnetic nanoparticles present in BCECs by targeted delivery to the transferrin receptor may undergo further transport into the brain when applying magnetic force. While magnetic immunoliposomes are targetable to BCECs, their design to enable further transport across the BBB when applying external magnetic force needs further improvement.
Sigma-Aldrich
Maleimide, 99%
Sigma-Aldrich
Didecyldimethylammonium bromide, 98%
Avanti
Soy PC, Avanti Research - A Croda Brand 840054C
Avanti
Soy PC, Avanti Research - A Croda Brand