Merck
  • Transforming Growth Factor-β1 Decreases β2-Agonist-induced Relaxation in Human Airway Smooth Muscle.
Transforming Growth Factor-β1 Decreases β2-Agonist-induced Relaxation in Human Airway Smooth Muscle.
American journal of respiratory cell and molecular biology (2019-02-12)
Christie A Ojiaku, Elena Chung, Vishal Parikh, Jazmean K Williams, Anthony Schwab, Ana Lucia Fuentes, Maia L Corpuz, Victoria Lui, Sam Paek, Natalia M Bexiga, Shreya Narayan, Francisco J Nunez, Kwangmi Ahn, Rennolds S Ostrom, Steven S An, Reynold A Panettieri
ABSTRACT
Helper T effector cytokines implicated in asthma modulate the contractility of human airway smooth muscle (HASM) cells. We have reported recently that a profibrotic cytokine, transforming growth factor (TGF)-β1, induces HASM cell shortening and airway hyperresponsiveness. Here, we assessed whether TGF-β1 affects the ability of HASM cells to relax in response to β2-agonists, a mainstay treatment for airway hyperresponsiveness in asthma. Overnight TGF-β1 treatment significantly impaired isoproterenol (ISO)-induced relaxation of carbachol-stimulated, isolated HASM cells. This single-cell mechanical hyporesponsiveness to ISO was corroborated by sustained increases in myosin light chain phosphorylation. In TGF-β1-treated HASM cells, ISO evoked markedly lower levels of intracellular cAMP. These attenuated cAMP levels were, in turn, restored with pharmacological and siRNA inhibition of phosphodiesterase 4 and Smad3, respectively. Most strikingly, TGF-β1 selectively induced phosphodiesterase 4D gene expression in HASM cells in a Smad2/3-dependent manner. Together, these data suggest that TGF-β1 decreases HASM cell β2-agonist relaxation responses by modulating intracellular cAMP levels via a Smad2/3-dependent mechanism. Our findings further define the mechanisms underlying β2-agonist hyporesponsiveness in asthma, and suggest TGF-β1 as a potential therapeutic target to decrease asthma exacerbations in severe and treatment-resistant asthma.
Sigma-Aldrich
Adenosine 3′,5′-cyclic monophosphate tris salt, ≥97% (HPLC), powder
Sigma-Aldrich
Anti-MLC-2 Antibody, clone 19D3.1, clone 19D3.1, from mouse