Merck
  • The late stage of COPI vesicle fission requires shorter forms of phosphatidic acid and diacylglycerol.
The late stage of COPI vesicle fission requires shorter forms of phosphatidic acid and diacylglycerol.
Nature communications (2019-08-01)
Seung-Yeol Park, Jia-Shu Yang, Zhen Li, Pan Deng, Xiaohong Zhu, David Young, Maria Ericsson, Ruben L H Andringa, Adriaan J Minnaard, Chunmei Zhu, Fei Sun, D Branch Moody, Andrew J Morris, Jun Fan, Victor W Hsu
ABSTRACT
Studies on vesicle formation by the Coat Protein I (COPI) complex have contributed to a basic understanding of how vesicular transport is initiated. Phosphatidic acid (PA) and diacylglycerol (DAG) have been found previously to be required for the fission stage of COPI vesicle formation. Here, we find that PA with varying lipid geometry can all promote early fission, but only PA with shortened acyl chains promotes late fission. Moreover, diacylglycerol (DAG) acts after PA in late fission, with this role of DAG also requiring shorter acyl chains. Further highlighting the importance of the short-chain lipid geometry for late fission, we find that shorter forms of PA and DAG promote the vesiculation ability of COPI fission factors. These findings advance a general understanding of how lipid geometry contributes to membrane deformation for vesicle fission, and also how proteins and lipids coordinate their actions in driving this process.
Product Description
Avanti
N-20:4 L-Serine (ARA-S), N-arachidonoyl L-serine, chloroform
Avanti
18:0-22:6 DG, 1-stearoyl-2-docosahexaenoyl-sn-glycerol, chloroform
Sigma-Aldrich
Magnesium acetate solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
L-α-Phosphatidylethanolamine, dioleoyl, ≥99% (GC), ≥98% (TLC), lyophilized powder
Avanti
18:0-22:6 PA, 1-stearoyl-2-docosahexaenoyl-sn-glycero-3-phosphate (sodium salt), chloroform
Avanti
10:0 PA, 1,2-didecanoyl-sn-glycero-3-phosphate (sodium salt), chloroform
Avanti
18:0 PA, Avanti Research - A Croda Brand
Avanti
10:0 PA, 1,2-didecanoyl-sn-glycero-3-phosphate (sodium salt), powder