Merck
  • Longitudinal Analysis Reveals Early Development of Three MPER-Directed Neutralizing Antibody Lineages from an HIV-1-Infected Individual.
Longitudinal Analysis Reveals Early Development of Three MPER-Directed Neutralizing Antibody Lineages from an HIV-1-Infected Individual.
Immunity (2019-03-17)
Shelly J Krebs, Young D Kwon, Chaim A Schramm, William H Law, Gina Donofrio, Kenneth H Zhou, Syna Gift, Vincent Dussupt, Ivelin S Georgiev, Sebastian Schätzle, Jonathan R McDaniel, Yen-Ting Lai, Mallika Sastry, Baoshan Zhang, Marissa C Jarosinski, Amy Ransier, Agnes L Chenine, Mangaiarkarasi Asokan, Robert T Bailer, Meera Bose, Alberto Cagigi, Evan M Cale, Gwo-Yu Chuang, Samuel Darko, Jefferson I Driscoll, Aliaksandr Druz, Jason Gorman, Farida Laboune, Mark K Louder, Krisha McKee, Letzibeth Mendez, M Anthony Moody, Anne Marie O'Sullivan, Christopher Owen, Dongjun Peng, Reda Rawi, Eric Sanders-Buell, Chen-Hsiang Shen, Andrea R Shiakolas, Tyler Stephens, Yaroslav Tsybovsky, Courtney Tucker, Raffaello Verardi, Keyun Wang, Jing Zhou, Tongqing Zhou, George Georgiou, S Munir Alam, Barton F Haynes, Morgane Rolland, Gary R Matyas, Victoria R Polonis, Adrian B McDermott, Daniel C Douek, Lawrence Shapiro, Sodsai Tovanabutra, Nelson L Michael, John R Mascola, Merlin L Robb, Peter D Kwong, Nicole A Doria-Rose
ABSTRACT
Lineage-based vaccine design is an attractive approach for eliciting broadly neutralizing antibodies (bNAbs) against HIV-1. However, most bNAb lineages studied to date have features indicative of unusual recombination and/or development. From an individual in the prospective RV217 cohort, we identified three lineages of bNAbs targeting the membrane-proximal external region (MPER) of the HIV-1 envelope. Antibodies RV217-VRC42.01, -VRC43.01, and -VRC46.01 used distinct modes of recognition and neutralized 96%, 62%, and 30%, respectively, of a 208-strain virus panel. All three lineages had modest levels of somatic hypermutation and normal antibody-loop lengths and were initiated by the founder virus MPER. The broadest lineage, VRC42, was similar to the known bNAb 4E10. A multimeric immunogen based on the founder MPER activated B cells bearing the unmutated common ancestor of VRC42, with modest maturation of early VRC42 intermediates imparting neutralization breadth. These features suggest that VRC42 may be a promising template for lineage-based vaccine design.
Product Description
Supelco
GC Stationary Phase, phase 1,2,3-tris(2-Cyanoethoxy)propane (TCEP), bottle of 50 g
Avanti
C12 Lactosyl(β) Ceramide (d18:1/12:0), Avanti Research - A Croda Brand 860545P, powder
Avanti
Ganglioside GM3 (Bovine Milk), Avanti Research - A Croda Brand
Sigma-Aldrich
Protease Inhibitor Cocktail powder, for general use, lyophilized powder
Sigma-Aldrich
1,2-Dimyristoyl-sn-glycero-3-phosphoethanolamine, synthetic, ≥99%
Avanti
C12 Glucosyl(β) Ceramide (d18:1/12:0), Avanti Research - A Croda Brand 860543P, powder
Avanti
14:0 PS, Avanti Research - A Croda Brand
Sigma-Aldrich
N-Stearoyl-D-sphingosine, ≥98.0% (TLC)