Merck
  • 25th anniversary article: MXenes: a new family of two-dimensional materials.
25th anniversary article: MXenes: a new family of two-dimensional materials.
Advanced materials (Deerfield Beach, Fla.) (2013-12-21)
Michael Naguib, Vadym N Mochalin, Michel W Barsoum, Yury Gogotsi
ABSTRACT
Recently a new, large family of two-dimensional (2D) early transition metal carbides and carbonitrides, called MXenes, was discovered. MXenes are produced by selective etching of the A element from the MAX phases, which are metallically conductive, layered solids connected by strong metallic, ionic, and covalent bonds, such as Ti2 AlC, Ti3 AlC2 , and Ta4 AlC3 . MXenes -combine the metallic conductivity of transition metal carbides with the hydrophilic nature of their hydroxyl or oxygen terminated surfaces. In essence, they behave as "conductive clays". This article reviews progress-both -experimental and theoretical-on their synthesis, structure, properties, intercalation, delamination, and potential applications. MXenes are expected to be good candidates for a host of applications. They have already shown promising performance in electrochemical energy storage systems. A detailed outlook for future research on MXenes is also presented.
Sigma-Aldrich
Titanium aluminium carbide 312, MAX Phase, ≥90%, ≤100 μm particle size
Sigma-Aldrich
Titanium aluminium carbide 312, MAX Phase, ≥90%, ≤40 μm particle size