- Characterization of a highly polymeric proanthocyanidin fraction from persimmon pulp with strong Chinese cobra PLA2 inhibition effects.
Characterization of a highly polymeric proanthocyanidin fraction from persimmon pulp with strong Chinese cobra PLA2 inhibition effects.
Fitoterapia (2011-11-01)
Shu-fen Xu, Bo Zou, Jie Yang, Ping Yao, Chun-mei Li
PMID22037569
ABSTRACT
Persimmon proanthocyanidin was fractionated on Toyopearl TSK-HW-50-F to yield a fraction with strong inhibition on the catalytic activity and edema-inducing activity and lethality of Chinese cobra PLA(2). Thiolysis suggested that the terminal units included C, EGCG and myricetin, and epicatechin, epigallocatechin, (epi)gallocatechin-3-O-gallate, and (epi)catechin-3-O-gallate occurred as extender units. The mean degree of polymerization was 23.7. MALDI TOF/MS, thioly-HPLC, FTIR and circular dichroism (CD) analyses showed that the fraction had high prodelphinidin content (55%) and a very high degree of 3-O-galloylation (92%). A type linkage is dominant in it and it had 4β linkage of the flavanyl substituent and 4R absolute configuration.
Product Description
TOYOPEARL® HW-50F Bulk Media, matrix hydroxylated methacrylate, bottle of 500 mL, 30-60 μm
TOYOPEARL® HW-65F Bulk Media, phase hydroxylated methacrylate, bottle of 500 mL, 30-60 μm particle size
TOYOPEARL® HW-55S Bulk Media, phase hydroxylated methacrylic polymer, bottle of 250 mL, 20-40 μm
Supelco
TOYOPEARL® HW-55F Bulk Media, phase hydroxylated methacrylate, bottle of 500 mL, 45 μm particle size
TOYOPEARL® HW-75F Bulk Media, phase hydroxylated methacrylate, bottle of 500 mL, 45 μm particle size
TOYOPEARL® HW-40S Bulk Media, phase hydroxylated methacrylate, bottle of 250 mL, 30 μm particle size
TOYOPEARL® HW-40F Bulk Media, phase hydroxylated methacrylate, bottle of 500 mL, 30-60 μm
TOYOPEARL® HW-40C Bulk Media, phase hydroxylated methacrylate, bottle of 500 mL, 75 μm particle size