Merck
An improved rapid method for the preparation of D-rhamnose.
Carbohydrate research (2011-11-22)
Olga N Smol'kina, Natalya S Shishonkova, Yulia P Fedonenko, Evelina L Zdorovenko, Svetlana A Konnova, Vladimir V Ignatov
ABSTRACT
A method is developed for the preparation of D-rhamnose from an O-polysaccharide (OPS) isolated by mild acid hydrolysis of Azospirillum brasilense SR75 cell mass. After the OPS hydrolysis, D-rhamnose was recovered by gel-permeation chromatography on Toyopearl TSK HW-40 and was crystallized. The sugar activity was demonstrated immunochemically. The advantages of the method are that it expedites and simplifies the extraction of D-rhamnose and increases its yield.
Product Description
TOYOPEARL® HW-50F Bulk Media, matrix hydroxylated methacrylate, bottle of 500 mL, 30-60 μm
TOYOPEARL® HW-65F Bulk Media, phase hydroxylated methacrylate, bottle of 500 mL, 30-60 μm particle size
TOYOPEARL® HW-55S Bulk Media, phase hydroxylated methacrylic polymer, bottle of 250 mL, 20-40 μm
Supelco
TOYOPEARL® HW-55F Bulk Media, phase hydroxylated methacrylate, bottle of 500 mL, 45 μm particle size
TOYOPEARL® HW-75F Bulk Media, phase hydroxylated methacrylate, bottle of 500 mL, 45 μm particle size
TOYOPEARL® HW-40S Bulk Media, phase hydroxylated methacrylate, bottle of 250 mL, 30 μm particle size
TOYOPEARL® HW-40F Bulk Media, phase hydroxylated methacrylate, bottle of 500 mL, 30-60 μm
TOYOPEARL® HW-40C Bulk Media, phase hydroxylated methacrylate, bottle of 500 mL, 75 μm particle size