Merck
  • Cloning, expression and characterization of the recombinant Yersinia pseudotuberculosis L-asparaginase.
Cloning, expression and characterization of the recombinant Yersinia pseudotuberculosis L-asparaginase.
Protein expression and purification (2012-01-10)
M V Pokrovskaya, S S Aleksandrova, V S Pokrovsky, N M Omeljanjuk, A A Borisova, N Yu Anisimova, N N Sokolov
ABSTRACT
We have cloned ansB (YPTB1411) gene from Yersinia pseudotuberculosis Q66CJ2 and constructed stable inducible expression system that overproduce L-asparaginase from Y. pseudotuberculosis (YpA) in Escherichiacoli BL21 (DE3) cells. For purification of YpA we used Q-Sepharose and DEAE-Toyopearl column chromatography. We examined kinetics of the enzyme reaction, catalytic activity as a function of pH, temperature and ionic strength, thermostability and other enzyme properties. Biochemical properties of YpA are similar with those of E. coli type II L-asparaginase. K(m) for L-asparagine is 17 ± 0.9 μM and pI 5.4 ± 0.3. Enzyme demonstrates maximum activity at pH 8.0 and 60 °C. YpA L-glutaminase activity is relatively low and more than 15 times less than specific activity towards L-asn. We evaluated also the antiproliferative effect of YpA in vitro and in vivo with E. colil-asparaginase (EcA) as the reference substance at similar conditions.
Product Description
TOYOPEARL® HW-50F Bulk Media, matrix hydroxylated methacrylate, bottle of 500 mL, 30-60 μm
TOYOPEARL® HW-65F Bulk Media, phase hydroxylated methacrylate, bottle of 500 mL, 30-60 μm particle size
TOYOPEARL® HW-55S Bulk Media, phase hydroxylated methacrylic polymer, bottle of 250 mL, 20-40 μm
Supelco
TOYOPEARL® HW-55F Bulk Media, phase hydroxylated methacrylate, bottle of 500 mL, 45 μm particle size
TOYOPEARL® HW-75F Bulk Media, phase hydroxylated methacrylate, bottle of 500 mL, 45 μm particle size
TOYOPEARL® HW-40S Bulk Media, phase hydroxylated methacrylate, bottle of 250 mL, 30 μm particle size
TOYOPEARL® HW-40F Bulk Media, phase hydroxylated methacrylate, bottle of 500 mL, 30-60 μm
TOYOPEARL® HW-40C Bulk Media, phase hydroxylated methacrylate, bottle of 500 mL, 75 μm particle size