Merck
  • Transcription Factor PROX1 Suppresses Notch Pathway Activation via the Nucleosome Remodeling and Deacetylase Complex in Colorectal Cancer Stem-like Cells.
Transcription Factor PROX1 Suppresses Notch Pathway Activation via the Nucleosome Remodeling and Deacetylase Complex in Colorectal Cancer Stem-like Cells.
Cancer research (2018-08-30)
Jenny Högström, Sarika Heino, Pauliina Kallio, Marianne Lähde, Veli-Matti Leppänen, Diego Balboa, Zoltán Wiener, Kari Alitalo
ABSTRACT
The homeobox transcription factor PROX1 is induced by high Wnt/β-catenin activity in intestinal adenomas and colorectal cancer, where it promotes tumor progression. Here we report that in LGR5+ colorectal cancer cells, PROX1 suppresses the Notch pathway, which is essential for cell fate in intestinal stem cells. Pharmacologic inhibition of Notch in ex vivo 3D organoid cultures from transgenic mouse intestinal adenoma models increased Prox1 expression and the number of PROX1-positive cells. Notch inhibition led to increased proliferation of the PROX1-positive colorectal cancer cells, but did not affect their ability to give rise to PROX1-negative secretory cells. Conversely, PROX1 deletion increased Notch target gene expression and NOTCH1 promoter activity, indicating reciprocal regulation between PROX1 and the Notch pathway in colorectal cancer. PROX1 interacted with the nucleosome remodeling and deacetylase (NuRD) complex to suppress the Notch pathway. Thus, our data suggests that PROX1 and Notch suppress each other and that PROX1-mediated suppression of Notch mediates its stem cell function in colorectal cancer.Significance: These findings address the role of the PROX1 homeobox factor as a downstream effector of Wnt/β-catenin singling in colorectal cancer stem cells and show that PROX1 inhibits the Notch pathway and helps to enforce the stem cell phenotype and inhibit differentiation. Cancer Res; 78(20); 5820-32. ©2018 AACR.
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, viscosity 2,600-5,600 cP, 2 % in H2O(20 °C)(lit.)
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Sigma-Aldrich
DAPT, ≥98% (HPLC), solid
Sigma-Aldrich
Biotin, ≥99% (HPLC), lyophilized powder
Sigma-Aldrich
Tamoxifen, ≥99%